Skip to Content
Merck
All Photos(1)

Key Documents

P1743

Sigma-Aldrich

Protein Phosphatase 2C from bovine brain

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$979.00

$979.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
$979.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$979.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

biological source

bovine brain

Quality Level

Assay

>90% (SDS-GE)

form

buffered aqueous glycerol solution

specific activity

~1000 units/mg protein

mol wt

42-45 kDa

packaging

vial of 1 μg

technique(s)

ligand binding assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

bovine ... PPP2CA(282320)

General description

Protein phosphatase 2C (PP2C) is a member of protein phosphatases, Mg2+/Mn2+ dependent (PPM) family. PP2C is a monomeric enzymes found in both prokaryotes and eukaryotes.[1]

Biochem/physiol Actions

In Arabidopsis, protein phosphatase 2C (PP2C) negatively regulates plant hormone, abscisic acid (ABA) signal transduction.[2][3] In eukaryotes, this protein modulates the cell cycle by reversing the activating phosphorylation of cyclin-dependent protein kinases (CDKs).[1] PP2C acts as a crucial physiological regulators of cell growth and of cellular stress signaling.[4]

Unit Definition

One unit will release 1.0 nanomole of phosphate from 32P-labeled myelin basic protein at pH 7.0 at 30 °C

Physical form

Solution in 50 mM Tris-HCl, pH 7.0, containing 14 mM 2-mercaptoethanol, 1 mM benzamidine, 0.1 mM PMSF, 1 mM EDTA, and 50% glycerol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of type 2C protein phosphatases in growth regulation and in cellular stress signaling.
Lammers T and Lavi S
Critical Reviews in Biochemistry and Molecular Biology, 42, 437-461 (2007)
ABI1 Protein Phosphatase 2C Is a Negative Regulator of Abscisic Acid Signaling
Gosti F, et al.
Plant Cell, 11, 1897-1910 (1999)
A protein phosphatase 2C involved in ABA signal transduction in Arabidopsis thaliana
Meyer K, et al.
Science, 264, 1452-1455 (1994)
Plant PP2C phosphatases: emerging functions in stress signaling.
Schweighofer A, et al.
Trends in Plant Science, 9, 236-243 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service