P0747
8-MA-cAMP
≥98% (HPLC), solid
Synonym(s):
8-Methylaminoadenosine-3′,5′-cyclic monophosphate sodium salt
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
solid
packaging
pkg of 10 μmol
color
white
solubility
H2O: freely soluble
shipped in
dry ice
storage temp.
−70°C
SMILES string
[Na+].CNc1nc2c(N)ncnc2n1[C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O
InChI
1S/C11H15N6O6P.Na/c1-13-11-16-5-8(12)14-3-15-9(5)17(11)10-6(18)7-4(22-10)2-21-24(19,20)23-7;/h3-4,6-7,10,18H,2H2,1H3,(H,13,16)(H,19,20)(H2,12,14,15);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1
InChI key
PYXGPPLPPUIPRE-MCDZGGTQSA-M
Biochem/physiol Actions
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service