Skip to Content
Merck
All Photos(1)

Key Documents

P0472

Sigma-Aldrich

Pilocarpine hydrochloride

meets USP testing specifications

Synonym(s):

(3S,4R)-4,5-Dihydro-3-ethyl-4-(1-methyl-1H-imidazol-5-ylmethyl)-2(3H)-furanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HCl
CAS Number:
Molecular Weight:
244.72
Beilstein:
4034491
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

form

solid

mp

202-205 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

Cl.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1

InChI key

RNAICSBVACLLGM-GNAZCLTHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maxime Lévesque et al.
Seizure, 25, 18-25 (2015-02-04)
Mesial temporal lobe epilepsy (MTLE) is the most prevalent type of partial epileptic disorders. In this study, we have analyzed the impact of levetiracetam (LEV) in the pilocarpine model of MTLE. Sprague-Dawley rats (n=19) were injected with pilocarpine (380 mg/kg
Manuela Mazzuferi et al.
Annals of neurology, 74(4), 560-568 (2013-05-21)
Epigenetic mechanisms involved in transcriptional regulation of multiple molecular pathways are potentially attractive therapeutic interventions for epilepsy, because single target therapies are unlikely to provide both anticonvulsant and disease-modifying effects. A selection of epilepsy-related gene expression data sets were retrieved
Kathleen Heng et al.
Epilepsia, 54(9), 1535-1541 (2013-07-16)
The role of granule cell axon (mossy fiber) sprouting in temporal lobe epileptogenesis is unclear and controversial. Rapamycin suppresses mossy fiber sprouting, but its reported effects on seizure frequency are mixed. The present study used high-dose rapamycin to more completely
Kevin D Phelan et al.
Molecular pharmacology, 83(2), 429-438 (2012-11-29)
Seizures are the manifestation of highly synchronized burst firing of a large population of cortical neurons. Epileptiform bursts with an underlying plateau potential in neurons are a cellular correlate of seizures. Emerging evidence suggests that the plateau potential is mediated
Jiandong Yu et al.
Journal of neurophysiology, 109(7), 1746-1763 (2013-01-18)
Temporal lobe epilepsy is associated with loss of interneurons and inhibitory dysfunction in the dentate gyrus. While status epilepticus (SE) leads to changes in granule cell inhibition, whether dentate basket cells critical for regulating granule cell feedforward and feedback inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service