Skip to Content
Merck
All Photos(2)

Key Documents

P0101

Sigma-Aldrich

Anti-Profilin 2 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PFN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~14 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

technique(s)

western blot: 1:500-1:1,000 using whole extracts of human SH-SY5Y cells, mouse brain, and rat brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PFN2(5217)
mouse ... Pfn2(18645)
rat ... Pfn2(81531)

General description

Profilins are actin-monomer-binding proteins, abundantly expressed in all eukaryotic cells from yeast to mammals. Profilin 2 (PFN2) is the neuronal specific isoform. It is an actin-binding protein and is a member of the small G-actin-binding class of protein.

Immunogen

synthetic peptide corresponding to amion acid residues 128-140 of human profilin 2 conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Application

Anti-Profilin 2 (C-terminal) antibody produced in rabbit has been used in immunostaining and immunoblotting.

Biochem/physiol Actions

Profilins are key regulators of actin dynamics. Profilin 2 (PFN2) associates with a number of proteins involved in membrane trafficking and endocytosis, such as synapsin and dynamin 1. It associates with dynamin 1 through the C-terminal proline-rich domain of dynamin 1 and thereby competes with the binding of SH3 ligands, such as endophilin and amphiphysin, thus interfering with the assembly of the endocytic machinery. Overexpression of profilin 2 inhibits endocytosis. Absence of profilin 2 in neurons increases endocytosis and membrane recycling. Profilin 2 is required for stimulated actin polymerization in the synapse and is involved in the regulation of synaptic vesicle exocytosis, neuronal excitability and complex behavior.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profilin2 contributes to synaptic vesicle exocytosis, neuronal excitability, and novelty-seeking behavior
Boyl PP, et al.
The Embo Journal, 26(12), 2991-2991 (2007)
Mouse profilin 2 regulates endocytosis and competes with SH3 ligand binding to dynamin 1
Gareus R, et al.
The Journal of biological chemistry, 281(5), 2803-2811 (2006)
Loss of profilin 2 contributes to enhanced epithelial-mesenchymal transition and metastasis of colorectal cancer
Zhang H, et al.
International Journal of Oncology, 53(3), 1118-1118 (2018)
Lisa Marie Walter et al.
Scientific reports, 11(1), 10287-10287 (2021-05-15)
Nuclear and cytoplasmic actin-cofilin rods are formed transiently under stress conditions to reduce actin filament turnover and ATP hydrolysis. The persistence of these structures has been implicated in disease pathology of several neurological disorders. Recently, the presence of actin rods
Lisa Marie Walter et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2147-2160 (2020-01-08)
Profilin is a major regulator of actin dynamics in multiple specific processes localized in different cellular compartments. This specificity is not only meditated by its binding to actin but also its interaction with phospholipids such as phosphatidylinositol (4,5)-bisphosphate (PIP2 )

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service