Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: Tac
Promoter activity: inducible
Promoter type: bacterial
Promoter activity: inducible
Promoter type: bacterial
Origin of replication:
pUC
Bacteria selection:
ampicillin
Peptide cleavage:
EKT
Pricing and availability is not currently available.
recombinant
expressed in E. coli
tag
FLAG® tagged
form
buffered aqueous solution
mol wt
size 5477 bp
bacteria selection
ampicillin
origin of replication
pUC
peptide cleavage
EKT
peptide tag location
N-terminal
promoter
Promoter name: Tac
Promoter activity: inducible
Promoter type: bacterial
shipped in
ambient
storage temp.
−20°C
General description
The pSF-TAC-NH2-FLAG expression vector is a 5.5 kb E. coli expression vector used for cloning and cytoplasmic expression of a properly inserted open reading frame as an N-terminal Met-FLAG fusion protein. The FLAG epitope is a small- hydrophilic- 8 amino acid-tag (DYKDDDDK) that enables sensitive detection and high quality protein purification using anti-FLAG products. The N-terminal Met-FLAG fusion protein may be detected using monoclonal anti-FLAG M1 (catalogue number F3040) or M2 (F3165) antibodies. The fusion protein may be purified using anti-FLAG affinity gel (A2220). In this vector, transcription is driven by the IPTG inducible TAC promoter.
This product is an alternative to our original pFLAG vector, product number E8083.
Application
Cloning in a gene: This plasmid contains a gene within the main multiple cloning site (NotI-ClaI). Any plasmid that we sell where the gene is this configuration will be located in the exact same position in relation to the start and stop codon of the gene. The only exceptions to this rule are fusions proteins where the fusion gene may be positioned at the front or end of the MCS to allow gene fusion.By positioning all of our genes in the same location it allows them to be transferred between plasmids using the same cloning method and restriction sites regardless of the plasmid being used from our product range. Inserting a new gene into this plasmid should be easily possible using a range of standard restriction enzyme sites that flank the gene currently in the vector.Multiple cloning site notes:In the multiple cloning site there are two important restriction sites called BsgI and BseRI sites. These sites both cut the DNA at the same position and cleave the stop codon of the gene in the multiple cloning site in this plasmid thereby producing a TA overhang. This overhang is compatible with any of our peptide or reporter fusion tag plasmids also cut with either of these enzymes. This allows seamless C-terminal fusions to be made with the gene in this multiple cloning site using a single cloning step from our C-terminal peptide and reporter tag product range. Normally the easiest method is to clone the C-terminal tag from our other plasmid products into this plasmid using BsgI or BseRI and the downstream ClaI restriction site.BseRI and BsgI sites are non-palindromic and cleave a defined number of bases away from their binding sites. This allows them to cut the upstream stop codon in the gene in this plasmid regardless of the gene sequence.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.oxgene.com
Other Notes
Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics™. Find out more at Oxford Genetics - Sigma′s partner for cloning and expression vectors for molecular biology and synthetic biology applications.
To view sequence information for this product, please visit the product page
Legal Information
Oxford Genetics is a trademark of Oxford Genetics Ltd
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Oxford Genetics is a trademark of Oxford Genetics Ltd
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Tecnologías de expresión de proteínas para varios sistemas de expresión que propician la investigación, la terapéutica y la producción de vacunas.
Protein expression technologies for various expression systems supporting research, therapeutics, and vaccine production.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| OGS632-5UG | 04061833351970 |