Skip to Content
Merck
All Photos(1)

Key Documents

O9505

Sigma-Aldrich

Orotidine

≥90% (HPLC)

Synonym(s):

Uridine-6-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O8
CAS Number:
Molecular Weight:
288.21
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥90% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C(=O)NC(=O)C=C2C(O)=O

InChI

1S/C10H12N2O8/c13-2-4-6(15)7(16)8(20-4)12-3(9(17)18)1-5(14)11-10(12)19/h1,4,6-8,13,15-16H,2H2,(H,17,18)(H,11,14,19)/t4-,6-,7-,8-/m1/s1

InChI key

FKCRAVPPBFWEJD-XVFCMESISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Orotidine, a nucleoside found naturally in fungi, bacteria and plants, may be used as a reference for the determination of presence of orotidine in urine or plasma due to urea cycle-related (transamidation) metabolic deficiencies or diseases.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Horiuchi et al.
Life sciences, 66(21), 2051-2070 (2000-05-24)
We investigated the relationship between the toxic effect of allopurinol and pyrimidine metabolism in mice. Allopurinol-induced increases in plasma transaminase levels in dinitrofluorobenzene (DNFB)-sensitized mice were not affected by uridine. In contrast, plasma creatinine and BUN tended to decrease 18
K J Kwon-Chung et al.
Journal of medical and veterinary mycology : bi-monthly publication of the International Society for Human and Animal Mycology, 30(1), 61-69 (1992-01-01)
Spontaneous mutants requiring uracil were isolated from both varieties of Cryptococcus neoformans by plating on 5-fluoroorotic acid (5-FOA) medium. Of the 36 strains tested (18 var. neoformans and 18 var. gattii), 24 (12 of each variety) generated 5-FOA-resistant cells requiring
Elizabeth M Brown et al.
PloS one, 8(3), e59237-e59237 (2013-03-28)
Analysis of the population genetic structure of microbial species is of fundamental importance to many scientific disciplines because it can identify cryptic species, reveal reproductive mode, and elucidate processes that contribute to pathogen evolution. Here, we examined the population genetic
A B Burlina et al.
Journal of inherited metabolic disease, 15(5), 707-712 (1992-01-01)
The allopurinol challenge test was performed on 44 healthy subjects (28 children and 16 adolescents) in order to establish normal values of urinary orotic acid excretion following allopurinol ingestion in the paediatric population. The subjects were divided into three groups
Annette Sievers et al.
Bioorganic chemistry, 33(1), 45-52 (2005-01-26)
The second order rate constant (k(cat)/K(m)) for decarboxylation of orotidine by yeast OMP decarboxylase (ODCase), measured by trapping (14)CO(2) released during the reaction, is 2 x 10(-4)M(-1)s(-1). This very low activity may be compared with a value of 3 x

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service