Skip to Content
Merck
All Photos(1)

Key Documents

O8890

Sigma-Aldrich

OU749

≥98% (HPLC)

Synonym(s):

N-[5-(4-Methoxybenzyl)-1,3,4-thiadiazol-2-yl]benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O3S2
CAS Number:
Molecular Weight:
361.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(Cc2nnc(NS(=O)(=O)c3ccccc3)s2)cc1

InChI

1S/C16H15N3O3S2/c1-22-13-9-7-12(8-10-13)11-15-17-18-16(23-15)19-24(20,21)14-5-3-2-4-6-14/h2-10H,11H2,1H3,(H,18,19)

InChI key

QCXWBMZVLJCKHF-UHFFFAOYSA-N

Biochem/physiol Actions

Gamma-glutamyl transpeptidase (GGT; aka gamma-glutamyl transferase) cleaves the gamma-glutamyl bond of glutathione, making extracellular glutathione available for intracellular use. Elevated intracellular glutathione contributes to resistance of tumors to chemotherapy and radiation. GGT is a therapeutic target for treatment-resistance cancers, as treatment with GGT inhibitors prior to chemotherapy or radiation could sensitize GGT-positive tumors to treatment by decreasing glutathione levels. Previously, all GGT inhibitors have been glutamine analogues, which are competitive for the gamma-glutamyl site. Due to high toxicity, these compounds cannot be used in the clinic. OU749 is the first non-glutamine GGT inhibitor. It is also non-competitive for the gamma-glutamyl site. OU749 inhibits human GGT in an enzyme assay and is much less toxic than other GGT inhibitors, including azaserine (glutamine analogue, Sigma catalog).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinlei Yu et al.
Scientific reports, 6, 30033-30033 (2016-07-19)
Although essential amino acids regulate mechanistic target of rapamycin complex 1 (mTORC1) and the integrated stress response (ISR), the role of cysteine is unknown. We found that in hepatoma HepG2 cells, cystine (oxidized form of cysteine) activated mTORC1 and suppressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service