Skip to Content
Merck
All Photos(2)

Documents

O8382

Sigma-Aldrich

17-Octadecynoic acid

≥95% (GC)

Synonym(s):

17-ODYA, Alkynyl Stearic Acid, Octadec-17-ynoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (GC)

form

powder

solubility

chloroform: 10 mg/mL to clear, colorless to faintly yellow

SMILES string

C#CCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h1H,3-17H2,(H,19,20)

InChI key

DZIILFGADWDKMF-UHFFFAOYSA-N

Application

17-Octadecynoic acid has been used in lipid synthesis.

Biochem/physiol Actions

17-Octadecynoic acid (7-ODYA) is an irreversible inhibitor of cytochrome P450 isozymes, that participates in long-chain fatty acid metabolism.
Suicide substrate inhibitor that selectively and irreversibly inhibits cytochrome P450 epoxygenases and ω-hydrolases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ossama A Hatoum et al.
American journal of physiology. Heart and circulatory physiology, 288(1), H48-H54 (2004-09-04)
The endothelium plays an important role in maintaining vascular homeostasis by synthesizing and releasing several mediators of vasodilation, which include prostacyclin (PGI(2)), nitric oxide, and endothelium-derived hyperpolarizing factor (EDHF). We have recently defined the role of nitric oxide and PGI(2)
Sphingosine-1-phosphate lyase deficient cells as a tool to study protein lipid interactions
Gerl M J, et al.
Testing, 11(4), e0153009-e0153009 (2016)
Effects of intrarenal infusion of 17-octadecynoic acid on renal antihypertensive mechanisms in anesthetized rabbits
Evans R G, et al.
American Journal of Hypertension, 11(7), 803-812 (1998)
M H Wang et al.
The Journal of pharmacology and experimental therapeutics, 284(3), 966-973 (1998-03-13)
We characterized the inhibitory activity of several acetylenic and olefinic compounds on cytochrome P450 (CYP)-derived arachidonic acid omega-hydroxylation and epoxidation using rat renal cortical microsomes and recombinant CYP proteins. Among the acetylenic compounds, 6-(2-propargyloxyphenyl)hexanoic acid (PPOH) and N-methylsulfonyl-6-(2-propargyloxyphenyl)hexanamide were found
Jacob S Yount et al.
Current protocols in chemical biology, 3(2), 65-79 (2011-05-01)
Protein fatty-acylation is the covalent addition of a lipid chain at specific amino acids. This modification changes the inherent hydrophobicity of a protein, often targeting it to cellular membrane compartments. Acylation may also regulate protein activity, stability, and protein-protein interactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service