Skip to Content
Merck
All Photos(2)

Key Documents

O6626

Sigma-Aldrich

3-(N,N-Dimethyloctylammonio)propanesulfonate inner salt

Synonym(s):

Octyl sulfobetaine, SB3-8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
279.44
Beilstein:
3962489
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Assay

≥98.0% (TLC)

form

powder

mol wt

279.44 g/mol

technique(s)

electrophoretic mobility shift assay: suitable
nucleic acid detection: suitable

conductivity

≤20 μmho per cm at 24 °C (0.1 M aqueous solution)

CMC

330 mM (20-25°C)

mp

212-215 °C (lit.)

SMILES string

CCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C13H29NO3S/c1-4-5-6-7-8-9-11-14(2,3)12-10-13-18(15,16)17/h4-13H2,1-3H3

InChI key

QZRAABPTWGFNIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(N,N-Dimethyloctylammonio)propanesulfonate is a zwitterionic detergent.

Application

3-(N,N-Dimethyloctylammonio)propanesulfonate has been used in a study to assess correlations between surrogate nitrogenous organic precursors and C-, N-DBP formation. [1] It has also been used in a study to investigate DNA binding site discrimination. [2] Zwitterionic detergent; mild solubilizer of plasma membranes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Satta et al.
Journal of chromatography, 299(1), 57-72 (1984-09-07)
A critical study has been made of the solubilizing properties of anionic, nonionic and zwitterionic surfactants to be used in the first dimension of two-dimensional isoelectric focusing-sodium dodecyl sulphate (IF-SDS) maps. Excess of SDS is a powerful solubilizing agent, but
Jill M Zalieckas et al.
Journal of bacteriology, 188(7), 2578-2585 (2006-03-21)
Two Bacillus subtilis transcriptional factors, TnrA and GlnR, regulate gene expression in response to changes in nitrogen availability. These two proteins have similar amino acid sequences in their DNA binding domains and bind to DNA sites (GlnR/TnrA sites) that have
Dickson Joseph et al.
Journal of colloid and interface science, 542, 308-316 (2019-02-15)
We report a one-pot seedless green method for the synthesis of gold-silver (AuAg) spiky branched nanostars, with gold (90%) being the major component. Here, the zwitterionic surfactant lauryl sulfobetaine (LSB) is employed in the synthesis of bimetallic nanostars. The concentration
H H Chang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 64(12), 2395-2403 (2011-12-16)
Nitrosamines have been emerging as disinfection byproducts in drinking water using source water impacted with domestic wastewaters. Nitrogenous organic compounds are suggested as precursors of nitrosamines, but many of them have not been identified. This study investigated the correlations between
Hyeok Jung Kim et al.
The Analyst, 144(7), 2248-2255 (2019-02-19)
Managing food contamination from bacteria has been an ongoing issue in the public health and industrial fields. Enzymatic substrates possessing optical properties, e.g. fluorescence or color manifestation, are widely exploited in pathogenic/non-pathogenic bacteria culture methods. Recently, various chromogenic substrates have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service