Skip to Content
Merck
All Photos(2)

Documents

O1883

Sigma-Aldrich

trans-13-Octadecenoic acid

~99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O2
CAS Number:
Molecular Weight:
282.46
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99% (GC)

storage temp.

−20°C

SMILES string

CCCC\C=C\CCCCCCCCCCCC(O)=O

Packaging

Sealed ampule under argon.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Emken et al.
Journal of lipid research, 24(1), 34-46 (1983-01-01)
The absorption and distribution of deuterated trans- and cis-13-octadecenoic acid (13t-18:1 and 13c-18:1) in plasma lipids were compared to deuterated cis-9-octadecenoic acid (9c-18:1) in two young adult male subjects. A mixture of triglycerides was fed in a multiple-labeled experiment where
Néstor M Carballeira et al.
Chemistry and physics of lipids, 161(1), 38-43 (2009-06-17)
The first total synthesis for the (Z)-17-methyl-13-octadecenoic acid was accomplished in seven steps and in a 45% overall yield. The use of (trimethylsilyl)acetylene was key in the synthesis. Based on a previous developed strategy in our laboratory the best synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service