Skip to Content
Merck
All Photos(1)

Key Documents

O1382

Sigma-Aldrich

Ochratoxin B

from Aspergillus ochraceus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO6
CAS Number:
Molecular Weight:
369.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Aspergillus ochraceus

storage temp.

2-8°C

SMILES string

CC1Cc2ccc(C(=O)NC(Cc3ccccc3)C(O)=O)c(O)c2C(=O)O1

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V G Curtui et al.
Food additives and contaminants, 18(8), 730-738 (2001-07-27)
Blood serum, kidney, liver and muscle sample per animal were collected from slaughtered pigs (n = 52). The samples were analysed for ochratoxin A (OTA) and B (OTB) by HPLC methods. Zearalenone (ZEA) in serum was analysed by enzyme immunoassay.
Mikiko Uchigashima et al.
Methods (San Diego, Calif.), 56(2), 180-185 (2011-09-29)
Two kinds of monoclonal antibodies (MoAbs), OCA-10A and OCA-1B, were prepared based on their specificity to ochratoxin A (OTA) and ochratoxin B (OTB) and on their tolerance to 40% methanol. In an indirect competitive enzyme-linked immunosorbent assay, the half maximal
Setsuko Tabata et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 49(2), 111-115 (2008-05-27)
Ochratoxin A (OTA), ochratoxin B (OTB) and citrinin (CIT) in commercial foods were simultaneously determined and confirmed with high-performance liquid chromatography (HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC/MS/MS). The samples examined were made up of cereal, fruit
Setsuko Tabata et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 49(2), 100-105 (2008-05-27)
Methods using high-performance liquid chromatography with fluorescence detection (HPLC-FL) and using liquid chromatography with tandem mass spectrometry (LC/MS/MS) were developed for simultaneous determination of ochratoxin A (OTA), ochratoxin B (OTB) and citrinin (CIT) in cereal, fruit, and coffee products. The
Günter Müller et al.
Toxicology, 199(2-3), 251-259 (2004-05-19)
Modulating effects of ochratoxin A (OTA) and some of its derivatives on viability and oxidative burst activity of porcine monocytes and granulocytes have been studied. The formation of free oxygen radicals by monocytes was suppressed by OTA and ochratoxin C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service