Skip to Content
Merck
All Photos(1)

Key Documents

O0766

Sigma-Aldrich

ONO-RS-082

≥97% (HPLC)

Synonym(s):

2-(p-Amylcinnamoyl)amino-4-chlorobenzoic acid, 4-Chloro-2-[[1-oxo-3-(4-pentylphenyl)-2-propen-1-yl]amino]-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClNO3
CAS Number:
Molecular Weight:
371.86
UNSPSC Code:
41106300
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

shelf life

Do not freeze

storage condition

protect from light

color

white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

Clc1cc(c(cc1)C(=O)O)NC(=O)\C=C\c2ccc(cc2)CCCCC

InChI

1S/C21H22ClNO3/c1-2-3-4-5-15-6-8-16(9-7-15)10-13-20(24)23-19-14-17(22)11-12-18(19)21(25)26/h6-14H,2-5H2,1H3,(H,23,24)(H,25,26)/b13-10+

InChI key

MDVFITMPFHDRBZ-JLHYYAGUSA-N

Biochem/physiol Actions

ONO-RS-082 is a reversible phospholipase A2 inhibitor.

Features and Benefits

This compound is featured on the Phospholipase A2 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rania Abu-Hamdah et al.
Cell biology international, 28(1), 7-17 (2004-02-05)
Aquaporins (AQP) are involved in rapid and active gating of water across biological membranes. The molecular regulation of AQP is unknown. Here we report the isolation, identification and reconstitution of the regulatory complex of AQP-1. AQP-1 and Galphai3 have been
Y Li et al.
American journal of physiology. Heart and circulatory physiology, 280(6), H2658-H2664 (2001-05-18)
Physiological concentrations of [Arg(8)]vasopressin (AVP; 10-500 pM) stimulate oscillations of cytosolic free Ca2+ concentration (Ca2+ spikes) in A7r5 vascular smooth muscle cells. We previously reported that this effect of AVP was blocked by a putative phospholipase A2 (PLA2) inhibitor, ONO-RS-082
H Yoshida et al.
The American journal of physiology, 273(3 Pt 1), G735-G747 (1997-10-08)
In an attempt to examine the structure-activity relationship of the cholecystokinin (CCK) peptide, we examined the structural motif of CCK truncated peptides responsible for rat pancreatic acinar amylase secretion and signal transduction. CCK-6 (Met28-Gly29-Trp30-Met31-Asp32-Phe33 -NH2), CCK-5 [CCK-(29-33)], and CCK-4 [CCK-(30-33)]
T Takeuchi et al.
Japanese journal of pharmacology, 87(4), 309-317 (2002-02-07)
The origin of Ca2+ necessary for carbachol (CCh)-induced contraction of longitudinal muscle of the proximal colon of rats was studied. CCh induced contraction of the muscle consisting of two phases, phasic and tonic phases, with a concomitant biphasic increase in
Y Sugita et al.
European journal of pharmacology, 365(1), 77-89 (1999-02-13)
In order to examine some possibly misleading conclusions of the pharmacological analysis of the signal transduction pathways of gastric acid secretion, we evaluated various agents including inhibitors of protein kinase C, cyclic AMP-dependent protein kinase, phospholipase C, phospholipase A2, lipoxygenase

Articles

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service