Skip to Content
Merck
All Photos(1)

Key Documents

O0641

Sigma-Aldrich

Oroxylin A

≥98% (HPLC)

Synonym(s):

5,7-Dihydroxy-6-methoxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-6-methoxyflavone;, 6-Methoxybaicalein;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

COc1c(O)cc2OC(=CC(=O)c2c1O)c3ccccc3

InChI

1S/C16H12O5/c1-20-16-11(18)8-13-14(15(16)19)10(17)7-12(21-13)9-5-3-2-4-6-9/h2-8,18-19H,1H3

InChI key

LKOJGSWUMISDOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oroxylin A is a flavonoid isolated from Scutellaria baicalensis, which is one of the most important medicinal herbs in traditional Korean/Chinese/Japanese medicine. It shows amelioration of Abeta(25-35) peptide-induced memory impairment.
Oroxylin A is a flavonoid isolated from Scutellaria baicalensis, which is one of the most important medicinal herbs in traditional Korean/Chinese/Japanese medicine. The amelioration of Abeta(25-35) peptide-induced memory impairment by oroxylin A is believed to be mediated via the GABAergic neurotransmitter system after a single administration, or by reductions in Abeta(25-35) peptide-induced astrocyte and microglia activations, iNOS expression, lipid peroxidation, and increased cholinergic neurotransmission after subchronic administration.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzu-Ling Tseng et al.
PloS one, 7(10), e47403-e47403 (2012-10-17)
Successful drug treatment for sepsis-related acute lung injury (ALI) remains a major clinical problem. This study was designed to assess the beneficial effects of post-treatment of oroxylin A (OroA), a flavonoid, in ameliorating lipopolysaccharides (LPS)-induced lung inflammation and fatality. Rats
Jingtian Qu et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 9257950-9257950 (2020-02-23)
Oroxylin A, a flavonoid, is naturally produced in many medicinal plants. Our previous study identified it as a phytoestrogen. Based on this, the present study investigated its vasoconstriction reducing effects and whether the action was mediated by the estrogen receptor
Hui-Ying Yang et al.
Molecular carcinogenesis, 51(2), 185-195 (2011-05-11)
In this study, oroxylin A showed strong reversal potency in BEL7402/5-FU cells and the reversal fold (RF) reached 4.69. Simultaneously, rhodamine-123 accumulation assay and flow cytometry analysis demonstrated oroxylin A could increase drug accumulation. When combined with oroxylin A, 5-FU
Runzhi Zhu et al.
PloS one, 8(8), e71612-e71612 (2013-08-21)
Based on the previous research that oroxylin A can suppress inflammation, we investigated the hepatoprotective role of oroxylin A against CCl₄-induced liver damage in mice and then studied the possible alteration of the activities of cytokine signaling participating in liver
Chenrui Li et al.
The AAPS journal, 14(1), 23-34 (2011-12-15)
Radix Scutellariae is a commonly used herbal medicine. Baicalein, wogonin, and oroxylin A are three major bioactive flavones in Radix Scutellariae and share similar chemical structures. The intestinal absorption and disposition of baicalein have been systematically investigated by our group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service