Skip to Content
Merck
All Photos(1)

Key Documents

O0266

Sigma-Aldrich

Obestatin mouse, rat

synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C114H174N34O31
CAS Number:
Molecular Weight:
2516.81
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic
form:
lyophilized powder
Assay:
≥97% (HPLC)

biological source

synthetic

Assay

≥97% (HPLC)

form

lyophilized powder

color

white

solubility

deionized water: 0.5-1 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H]2CCCN2C(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](N)Cc3ccccc3)C(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc5c[nH]cn5)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(N)=O

InChI

1S/C114H174N34O31/c1-12-60(8)93(146-90(157)54-128-111(177)92(59(6)7)147-109(175)81(50-91(158)159)143-108(174)78(46-65-25-17-14-18-26-65)144-110(176)83-29-22-42-148(83)113(179)63(11)132-105(171)80(49-87(120)154)139-97(163)69(116)45-64-23-15-13-16-24-64)112(178)137-71(27-19-20-40-115)101(167)140-76(44-58(4)5)106(172)145-82(55-149)99(165)127-52-88(155)130-61(9)95(161)134-72(34-37-84(117)151)103(169)141-77(47-66-30-32-68(150)33-31-66)107(173)136-73(35-38-85(118)152)102(168)135-74(36-39-86(119)153)104(170)142-79(48-67-51-124-56-129-67)98(164)126-53-89(156)133-70(28-21-41-125-114(122)123)100(166)131-62(10)96(162)138-75(94(121)160)43-57(2)3/h13-18,23-26,30-33,51,56-63,69-83,92-93,149-150H,12,19-22,27-29,34-50,52-55,115-116H2,1-11H3,(H2,117,151)(H2,118,152)(H2,119,153)(H2,120,154)(H2,121,160)(H,124,129)(H,126,164)(H,127,165)(H,128,177)(H,130,155)(H,131,166)(H,132,171)(H,133,156)(H,134,161)(H,135,168)(H,136,173)(H,137,178)(H,138,162)(H,139,163)(H,140,167)(H,141,169)(H,142,170)(H,143,174)(H,144,176)(H,145,172)(H,146,157)(H,147,175)(H,158,159)(H4,122,123,125)/t60-,61-,62-,63-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,92-,93-/m0/s1

InChI key

OJSXICLEROKMBP-FFUDWAICSA-N

Gene Information

mouse ... Ghrl(58991)
rat ... Ghrl(59301)

Amino Acid Sequence

Phe-Asn-Ala-Pro-Phe-Asp-Val-Gly-Ile-Lys-Leu-Ser-Gly-Ala-Gln-Tyr-Gln-Gln-His-Gly-Arg-Ala-Leu-NH2

General description

Obestatin is a 23 amino acid bioactive peptide encoded by the ghrelin gene.

Application

Obestatin has been used to study its effect on serum glucose and enzymatic parameters in rats and used to investigate its effect on plasma acyl-ghrelin and body weight in wistar rat.

Biochem/physiol Actions

Obestatin opposes the action of circulating appetite-inducing hormone-ghrelin and plays a vital role in weight regulation via hindering food intake, jejunal contraction and reducing the body weight. Obestatin mediates its activity by interacting with the G-protein coupled receptor 39 (GPR-39).
Obestatin, an obesity peptide, along with Ghrelin peptides are derived from the same prepropeptide, Ghrelin. In contrast to Ghrelin, Obestatin suppresses food intake, body weight gain, and gastrointestinal motility in rodents.

Physical form

Lyophilized from 0.1% aqueous trifluoroacetate (TFA).

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of short- and long-term treadmill exercises on levels of ghrelin, obestatin and NPY in plasma and brain extraction of obese rats.
Wang J
Endocrine, 33(1), 77-83 (2008)
Effect of Peripheral Obestatin Infusion on Serum Glucose and some Enzymatic Parameters in rats
Rad M N
Advances in Bioresearch, 7 (3), 38-41 (2016)
Obestatin, a peptide encoded by the ghrelin gene, opposes ghrelin's effects on food intake.
Zhang JV
Science, 310(5750), 996-999 (2005)
Eman F Khaleel et al.
Archives of physiology and biochemistry, 125(1), 64-78 (2018-02-13)
This study investigated the ameliorative and protective effects of long-term obestatin administration (80 nmol/kg/ intraperitoneal injection (i.p.)) on the pathogenesis of high-fat diet (HFD) induced nonalcoholic fatty liver disease (NAFLD) in rats. Rats (n = 8/group) were divided as control, NAFLD, NAFLD + Simvastatin, NAFLD + obestatin
Effects of obestatin on energy balance and growth hormone secretion in rodents.
Nogueiras R
Endocrinology, 148(1), 21-26 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service