Skip to Content
Merck

NA2000

GenElute Blood Genomic DNA Kit

sufficient for 10 purifications

Synonym(s):

Gen Elute

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 10 purifications

General description

The GenElute Blood Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from fresh or aged (older than 24 hours) whole blood. The kit combines the advantages of silica binding with a microspin format, and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

The purified genomic DNA is ready for downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • sequencing reactions
  • cloning

Biochem/physiol Actions

The starting material is lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes the DNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. A Prewash Solution is provided to help remove contaminants that are associated with aged (older than 24 hours) whole blood samples. After washing to remove contaminants, the DNA is eluted in 200 mL of a Tris-EDTA solution.

The expected yields of genomic DNA will vary depending on the amount and nature of the starting material used (for example, 4 to 10 μg of RNase A-treated DNA can be isolated from 200 μl of fresh whole blood in less than one hour). DNA purified with this kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Features and Benefits

  • Starting material: Up to 200 μl of fresh or aged blood
  • Expected yield: Up to 10 μg
  • Elution volume: 400 μl
  • Time required: <40 min
  • A260/A280 ratio: 1.6 - 1.9
  • Compatible with many anticoagulants, including EDTA, Heparin, and Sodium Citrate
  • Typical yields 4-10 μg from 200 μL of whole blood
  • Pure genomic DNA with a A260/A280 ratio between 1.6 and 1.9
  • Isolate DNA up to 50 kb in length

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under GenElute Blood Genomic DNA Kit




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library