Skip to Content
Merck
All Photos(7)

Key Documents

NA0200S

Sigma-Aldrich

GenElute HP Plasmid Midiprep Kit

sufficient for 4 purifications

Synonym(s):

GenElute HP Plasmid Midiprep Kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

Pricing and availability is not currently available.

usage

sufficient for 4 purifications

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

storage temp.

15-25°C

General description

The GenElute HP Plasmid Midiprep Purification kits offer an ultrafast and efficient solution for large-scale plasmid preparation from E. coli cultures. These kits combine silica-based membrane technology and the convenience of spin or vacuum format. The kits also include a special filter cartridge, which replaces the centrifugation step following alkaline lysis. Following lysis the DNA is bound to the silica membrane and contaminants are removed with a simple wash step. The DNA is then eluted in water or TE buffer by centrifugation.

The purified plasmid DNA is suitable for a wide variety of molecular biology applications, including restriction digestions and sequencing.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Features and Benefits

  • From harvested bacterial culture to pure plasmid DNA in 30 minutes or less
  • Up to 350 μg of high-copy plasmid DNA
  • Flexibility of a vacuum or spin format
  • No phenol/chloroform extraction or alcohol precipitation required
  • Contains fewer plastic components, reducing the amount of waste

Other Notes

For additional information, please see www.sigma-aldrich.com/genelutehp.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service