Skip to Content
Merck
All Photos(5)

Key Documents

N8264

Sigma-Aldrich

Nucleoside Phosphorylase from microorganisms

lyophilized powder, ≥10 units/mg protein

Synonym(s):

Nucleoside Phosphorylase bacterial, PNP, Purine nucleoside phosphorylase, Purine nucleoside:orthophosphate ribosyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

form

lyophilized powder

specific activity

≥10 units/mg protein

mol wt

~120 kDa

composition

Protein, 40-80%

storage temp.

−20°C

SMILES string

[n]2(c3ncncc3nc2)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C10H12N4O4/c15-2-6-7(16)8(17)10(18-6)14-4-13-5-1-11-3-12-9(5)14/h1,3-4,6-8,10,15-17H,2H2/t6-,7-,8-,10-/m1/s1

InChI key

MRWXACSTFXYYMV-FDDDBJFASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nucleoside Phosphorylase are trimeric and have a molecular mass of about 100 kDa.[1] Each monomer consists of a mixed β-sheet core flanked by eight α-helices and a purine binding in hydrophobic pocket.[1]
Nucleoside phosphorylase is an enzyme important for the biosynthesis of nucleosides. [2]

Application

Nucleoside Phosphorylase from microorganisms has been used:
  • in phosphopentomutase assay for adenosine synthesis[3]
  • for the conversion of 7-methylthioguanosine to 7-methylthioguanine[4][5]
  • in the synthesis of cytokinins in lyophilized cotton plant samples[6]

Nucleoside phosphorylase is used in coupled enzyme systems to measure protein dephosphorylation.

Biochem/physiol Actions

Catalyzes the following reaction:purine nucleoside + phosphate = purine + alpha-D-ribose 1-phosphate
Nucleoside Phosphorylase catalyzes the synthesis of nucleoside derivatives.[7]
This enzyme is useful for enzymatic determination of inorganic phosphorus, 5′-nucleotidase and adenosine deaminase when coupled with xanthine oxidase (XTO-212) and uricase (UAO-201, UAO-211). Purine nucleoside phosophorylase has shown the ability to perform both phosphorylosis and synthesis of purine deoxy- and ribonucleosides. [8] It has also been found that membrane-associated nucleoside phosphorylases may have a transmembranal orientation with their base and ribose-1-P binding sites on opposite sides of the membrane. [9]

Physical properties

Isoelectric point : 4.1 +/- 0.1
Michaelis constants : 6.4 x 10-5M (Inosine), 3.2x10-4M (Pi)
Inhibitors : p-Chloromercuribenzoate, SDS, Hg++, Ag+Optimum pH : 7.5 - 8.0
Optimum temperature : 65oC
pH Stability : pH 6.0 - 9.0 (30oC, 16hr)
Thermal stability : below 60oC (pH 7.7, 30min)

Unit Definition

One unit will cause the phosphorolysis of 1.0 μmole of inosine to hypoxanthine and ribose 1-phosphate per min at pH 7.4 at 25 °C.

Physical form

Lyophilized powder containing potassium gluconate, mannitol and EDTA

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanan M A Moustafa et al.
Analytical biochemistry, 501, 75-81 (2016-03-01)
Phosphopentomutase (PPM) catalyzes the interconversion of α-D-(deoxy)-ribose 1-phosphate and α-D-(deoxy)-ribose 5-phosphate. We developed a coupled or uncoupled enzymatic assay with an enzyme nucleoside phosphorylase for determining PPM activities on D-ribose 5-phosphate at a broad temperature range from 30 to 90
E C Keystone et al.
Annals of the rheumatic diseases, 68(6), 789-796 (2008-12-11)
The phase III GO-FORWARD study examined the efficacy and safety of golimumab in patients with active rheumatoid arthritis (RA) despite methotrexate therapy. Patients were randomly assigned in a 3 : 3 : 2 : 2 ratio to receive placebo injections
Enzymatic synthesis of nucleosides by nucleoside phosphorylase co-expressed in Escherichia coli
Ding, Q., et al.
Journal of Zhejiang University. Science, 11, 880-888 (2010)
A second purine nucleoside phosphorylase in Escherichia coli K-12. II. Properties of xanthosine phosphorylase and its induction by xanthosine
Hmmer-Jespersen, K.
Molecular Genetics and Genomics, 179, 341-348 (1980)
Measurement of nonribosomal peptide Synthetase Adenylation domain activity using a continuous hydroxylamine release assay
Duckworth BP, et al.
Methods in Molecular Biology, 53-61 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service