N7002
DL-Norleucine methyl ester hydrochloride
≥98% (TLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H15NO2 · HCl
CAS Number:
Molecular Weight:
181.66
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
Product Name
DL-Norleucine methyl ester hydrochloride,
Assay
≥98% (TLC)
form
powder
color
white to off-white
application(s)
detection
SMILES string
Cl.CCCCC(N)C(=O)OC
InChI
1S/C7H15NO2.ClH/c1-3-4-5-6(8)7(9)10-2;/h6H,3-5,8H2,1-2H3;1H
InChI key
FMMOVZRXLNVNBI-UHFFFAOYSA-N
Biochem/physiol Actions
DL-Norleucine methyl ester hydrochloride is a neurotransmitter commonly used as an internal standard.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kumiko Sakai-Kato et al.
Analytical chemistry, 74(13), 2943-2949 (2002-07-27)
A novel trypsin-encapsulation technique using the sol-gel method was developed for the preparation of an on-line enzyme reactor integrated into capillary electrophoresis. Trypsin was encapsulated in tetramethoxysilane-based hydrogel, and its enzymatic activity was evaluated using alpha-N-benzoyl-L-arginine ethyl ester and two
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service