Skip to Content
Merck
All Photos(1)

Key Documents

N6127

Sigma-Aldrich

19-Norethindrone acetate

Synonym(s):

17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one 17-acetate, 17α-Ethynyl-19-nortestosterone 17β-acetate, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one acetate, 19-Norethisterone 17-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

Assay

≥98.00% (HPLC)

form

powder

solubility

chloroform: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C

InChI

1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,13,17-20H,5-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1

InChI key

IMONTRJLAWHYGT-ZCPXKWAGSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eija Pöllänen et al.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 20(5), 372-379 (2010-08-21)
To investigate the effects of combined hormone replacement therapy (HRT) and its effective agents on the IGF-1 signaling pathway. To examine the effects of HRT on skeletal muscle in vivo, we utilized pre- and post-intervention samples from a randomized double
Anieta M Sieuwerts et al.
Molecular oncology, 5(6), 504-516 (2011-10-01)
Risk assessment of future breast cancer risk through exposure to sex steroids currently relies on clinical scorings such as mammographic density. Knowledge about the gene expression patterns in existing breast cancer tumors may be used to identify risk factors in
D J Kaser et al.
Journal of pediatric and adolescent gynecology, 25(2), 105-108 (2011-12-14)
To evaluate the efficacy and tolerability of norethindrone acetate (NA) as single-agent hormonal therapy for suppression of endometriosis symptoms in adolescents and young adults. Retrospective study. Two academic medical centers. A keyword search using the query 'NA' was applied to
Marjo Tuppurainen et al.
Maturitas, 66(4), 423-430 (2010-06-16)
To determine the effects of HRT with or without clodronate on bone mineral density (BMD) change and bone turnover markers. Prospective, partly randomized trial. Kuopio University Hospital, Finland. 167 osteoporotic women (61+/-2.7 years; T-score<or=-2.5 SD). Estradiol 2 mg+NETA 1 mg
Donita Africander et al.
Contraception, 84(4), 423-435 (2011-09-17)
Medroxyprogesterone acetate (MPA) and norethisterone (NET) and its derivatives are widely used in female reproductive therapy, but little is known about their mechanisms of action via steroid receptors in the female genital tract. MPA used as a contraceptive has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service