Skip to Content
Merck
All Photos(1)

Key Documents

N5912

Sigma-Aldrich

Anti-NORE1 (RASSF5) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RAPL, Anti-Ras Association (RalGDS/AF-6) Domain Family 5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASSF5(83593)
mouse ... Rassf5(54354)
rat ... Rassf5(54355)

General description

Ras association domain family member 5 (RASSF5) is also known as novel Ras effector- 1 (NORE-1). It exists in three isoforms and the gene encoding it is located on human chromosome 1q32.1.

Immunogen

peptide corresponding to the NORE1 protein (amino acids 313-329). This sequence is located in the RA domain of NORE1 and is 100% conserved between human, mouse, rat, and hamster.

Biochem/physiol Actions

Ras association domain family member 5 (RASSF5) is a tumor suppressor protein. It interacts with kinases like mammalian sterile 20-like kinase 1/2 (MST1/2). RASSF5 is involved in the regulation of cell cycle and apoptosis.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu-Hui Zhou et al.
Oncology reports, 32(4), 1505-1512 (2014-08-12)
Ras association (RalGDS/AF-6) domain family member RASSF5 has been implicated in a variety of key biological processes, including cell proliferation, cell cycle regulation and apoptosis. It is believed to play an important role in tumorigenesis as a tumor suppressor in
Katherine K Slemmons et al.
Cancer research, 80(14), 3046-3056 (2020-05-02)
Rhabdomyosarcoma is the most common childhood soft-tissue sarcoma, yet patients with metastatic or recurrent disease continue to do poorly, indicating a need for new treatments. The SRC family tyrosine kinase YES1 is upregulated in rhabdomyosarcoma and is necessary for growth
Hye Jin Kim et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 68(Pt 7), 813-815 (2012-07-04)
NORE1 is an important tumour suppressor in human cancers that interacts with the pro-apoptotic protein kinase MST1/2 through SARAH domains. The SARAH domain (residues 366-413) of human NORE1 was expressed in Escherichia coli, purified and crystallized using the hanging-drop vapour-diffusion
Xiaoli Liu et al.
Frontiers in oncology, 10, 600927-600927 (2021-01-26)
The principal issue derived from thyroid cancer is its high propensity to metastasize to the lymph node. Aberrant exprssion of long non-coding RNAs have been extensively reported to be significantly correlated with lymphatic metastasis of thyroid cancer. However, the clinical
Wei Guo et al.
Clinical & experimental metastasis, 32(1), 83-98 (2015-01-13)
As a result of alternative splicing and differential promoter usage, RASSF5 exists in at least three isoforms (RASSF5A-RASSF5C), which may play different roles in tumorigenesis. The present study was to detect the role of RASSF5A, B and C in esophageal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service