Skip to Content
Merck
All Photos(1)

Documents

N5023

Sigma-Aldrich

Nordihydroguaiaretic acid

≥90% (HPLC), from Larrea divaricata (creosote bush)

Synonym(s):

1,4-Bis(3,4-dihydroxyphenyl)-2,3-dimethylbutane, 4,4′-(2,3-Dimethyltetramethylene)dipyrocatechol, Masoprocol, NDGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(CH3)CH2C6H3-1,2-(OH)2]2
CAS Number:
Molecular Weight:
302.36
Beilstein:
2056826
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Larrea divaricata (creosote bush)

Assay

≥90% (HPLC)

form

crystalline

mp

184-186 °C (lit.)

solubility

ethanol: 50 mg/mL, clear to slightly hazy

SMILES string

CC(Cc1ccc(O)c(O)c1)C(C)Cc2ccc(O)c(O)c2

InChI

1S/C18H22O4/c1-11(7-13-3-5-15(19)17(21)9-13)12(2)8-14-4-6-16(20)18(22)10-14/h3-6,9-12,19-22H,7-8H2,1-2H3

InChI key

HCZKYJDFEPMADG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lipoxygenase inhibitor; polyphenol-bearing o-dihydroxy (catechol) structure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena Palacio et al.
Plant science : an international journal of experimental plant biology, 193-194, 1-7 (2012-07-17)
The lignan nordihydroguaiaretic acid (NDGA) and its derivatives existing in Larrea divaricata species show a wide range of pharmacological activities which makes this genus an interesting target to consider the plant in vitro cultivation systems as a feasible alternative source
Silvia Arteaga et al.
Journal of ethnopharmacology, 98(3), 231-239 (2005-04-09)
Although controversial, Creosote bush, Larrea tridentata (Sesse and Moc. ex DC) Coville, is used to treat a variety of illnesses including infertility, rheumatism, arthritis, diabetes, gallbladder and kidney stones, pain and inflammation. Recently, it has been used as a nutritional
Victor Kenyon et al.
Journal of medicinal chemistry, 54(15), 5485-5497 (2011-07-12)
We report the discovery of novel small molecule inhibitors of platelet-type 12-human lipoxygenase, which display nanomolar activity against the purified enzyme, using a quantitative high-throughput screen (qHTS) on a library of 153607 compounds. These compounds also exhibit excellent specificity, >50-fold
Nassira Mahmoudi et al.
Antimicrobial agents and chemotherapy, 52(4), 1215-1220 (2008-01-24)
We conducted a quantitative structure-activity relationship (QSAR) study based on a database of 127 compounds previously tested against the liver stage of Plasmodium yoelii in order to develop a model capable of predicting the in vitro antimalarial activities of new
Judith Sylvester et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 30(3), 563-574 (2012-07-27)
We investigated the unknown molecular mechanisms of Interleukin-1 (IL-1β)-induced cartilage aggrecan degeneration by aggrecanase (ADAMTS-A Disintegrin And Metalloproteinase with ThromboSpondin motifs) in human articular chondrocytes, a model mimicking human arthritis. Chondrocytes were pretreated with various pharmacological inhibitors and then stimulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service