N3529
NAN-190 hydrobromide
Synonym(s):
1-(2-Methoxyphenyl)-4-[4-(2-phthalimido)butyl]piperazine hydrobromide
About This Item
Recommended Products
SMILES string
Br.COc1ccccc1N2CCN(CCCCN3C(=O)c4ccccc4C3=O)CC2
InChI
1S/C23H27N3O3.BrH/c1-29-21-11-5-4-10-20(21)25-16-14-24(15-17-25)12-6-7-13-26-22(27)18-8-2-3-9-19(18)23(26)28;/h2-5,8-11H,6-7,12-17H2,1H3;1H
InChI key
AXRUEPFPTQYHQD-UHFFFAOYSA-N
Gene Information
human ... HTR1A(3350)
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service