Skip to Content
Merck
All Photos(1)

Key Documents

N3262

SAFC

NCTC 135 Medium

With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.75

form

powder

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: no
L-glutamine: 0.13573 g/L
glucose: 1.0 g/L
phenol red: 0.02 g/L

shipped in

ambient

storage temp.

2-8°C

General description

NCTC 135 Medium is ideal for serum-free growth of mouse L-cells.

Application

NCTC 135 Medium has been used:
  • to culture Philophthalmus sp. rediae in vitro
  • to culture Ehrlich ascites mouse tumor cell line
  • as a culture media for the differentiation of Rcho-1 cells

Quantity

Formulated to contain 9.3 grams of powder per liter of medium.

Reconstitution

Supplement with 2.2 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Melanie M Lloyd et al.
Experimental parasitology, 129(2), 101-106 (2011-08-02)
The ability to culture parasites outside their host (i.e. in vitro) is essential for several aspects of parasitological research. Here, a culture medium for marine trematode parthenitae was optimized using Philophthalmus sp. rediae from the intermediate snail host, Zeacumantus subcarinatus.
Synthesis, characterization and antitumor activity of 1, 2-disubstituted ferrocenes and cyclodextrin inclusion complexes
Petrovski Z, et al.
Journal of Organometallic Chemistry, 693, 675-684 (2008)
Aiko Aoki et al.
The American journal of pathology, 188(11), 2474-2486 (2018-08-31)
Hypertensive disorder of pregnancy (HDP) is a serious pregnancy complication that is life threatening to both the mother and fetus. Understanding HDP pathophysiology is important for developing medical treatments. This study demonstrates the involvement of autophagy deficiency in adverse maternal
Melanie M Lloyd et al.
Experimental parasitology, 129(2), 101-106 (2011-08-02)
The ability to culture parasites outside their host (i.e. in vitro) is essential for several aspects of parasitological research. Here, a culture medium for marine trematode parthenitae was optimized using Philophthalmus sp. rediae from the intermediate snail host, Zeacumantus subcarinatus.
Aiko Aoki et al.
The American journal of pathology, 188(11), 2474-2486 (2018-08-31)
Hypertensive disorder of pregnancy (HDP) is a serious pregnancy complication that is life threatening to both the mother and fetus. Understanding HDP pathophysiology is important for developing medical treatments. This study demonstrates the involvement of autophagy deficiency in adverse maternal

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Related Content

NCTC 109 Medium Formulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service