Skip to Content
Merck
All Photos(1)

Key Documents

N3253

Sigma-Aldrich

2-Nitrophenyl β-D-fucopyranoside

≥99% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO7
CAS Number:
Molecular Weight:
285.25
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥99% (TLC)

storage temp.

−20°C

SMILES string

CC1OC(Oc2ccccc2N(=O)=O)C(O)C(O)C1O

InChI

1S/C12H15NO7/c1-6-9(14)10(15)11(16)12(19-6)20-8-5-3-2-4-7(8)13(17)18/h2-6,9-12,14-16H,1H3

InChI key

SWRPIVXPHLYETN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Bell et al.
Nature structural biology, 7(3), 209-214 (2000-03-04)
Crystal structures of the Lac repressor, with and without isopropyithiogalactoside (IPTG), and the repressor bound to operator have provided a model for how the binding of the inducer reduces the affinity of the repressor for the operator. However, because of
S P Manly et al.
Biochemistry, 24(15), 3842-3846 (1985-07-16)
The thermal denaturation of the core protein of lac repressor was studied alone and in the presence of the inducer isopropyl beta-D-thiogalactoside (IPTG) and the antiinducer o-nitrophenyl beta-D-fucoside (ONPF) by means of high-sensitivity differential scanning calorimetry. The denaturation that takes
Corey J Wilson et al.
Biophysical chemistry, 126(1-3), 94-105 (2006-07-25)
Specific interactions between proteins and ligands that modify their functions are crucial in biology. Here, we examine sugars that bind the lactose repressor protein (LacI) and modify repressor affinity for operator DNA using isothermal titration calorimetry and equilibrium DNA binding
N Horton et al.
Journal of molecular biology, 265(1), 1-7 (1997-01-10)
The wild type E. coli lac operator is embedded in a 35 base-pair DNA sequence containing extensive 2-fold symmetry, suggesting a symmetric repressor operator complex. However, deviations from strict 2-fold symmetry occur at the central base-pair and at three additional
Liskin Swint-Kruse
Biochemistry, 43(34), 10886-10895 (2004-08-25)
The vast increase in available data from the "-omics" revolution has enabled the fields of structural proteomics and structure prediction to make great progress in assigning realistic three-dimensional structures to each protein molecule. The challenge now lies in determining the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service