Skip to Content
Merck
All Photos(2)

Key Documents

N3136

Sigma-Aldrich

Naltrexone hydrochloride

≥99% (HPLC), powder, opioid receptor antagonist

Synonym(s):

Naltrexone HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO4 · HCl
CAS Number:
Molecular Weight:
377.86
Beilstein:
3580333
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Naltrexone hydrochloride,

form

powder

originator

Novartis

storage temp.

2-8°C

SMILES string

Cl.Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O)CC6CC6

InChI

1S/C20H23NO4.ClH/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11;/h3-4,11,15,18,22,24H,1-2,5-10H2;1H/t15-,18+,19+,20-;/m1./s1

InChI key

RHBRMCOKKKZVRY-ITLPAZOVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Naltrexone hydrochloride has been used:
  • as an opioid antagonist, to analyse its effect on ethanol preference using Caenorhabditis elegans as a model[1]
  • to determine its effectiveness in reducing the preference for substance of abuse (SOA) like nicotine and cocaine using Caenorhabditis elegans as a model[2]
  • in the preparation of combinatorial drug, PXT3003 for treating Charcot-Marie-Tooth disease 1A (CMT1A) transgenic rat model Pmp22[3]

Biochem/physiol Actions

Competitive antagonist for μ, κ, δ, and σ-opioid receptors; has greater oral efficacy and longer duration of action than naloxone.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Moreno-Vicente et al.
Journal of pharmaceutical and biomedical analysis, 114, 105-112 (2015-06-04)
A bioanalytical method using high-performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) was developed and validated for simultaneous quantification of heroin, its main metabolites and naloxone. In addition, naltrexone was detected qualitatively. This method was used to analyse human plasma samples from
Early short-term PXT3003 combinational therapy delays disease onset in a transgenic rat model of Charcot-Marie-Tooth disease 1A (CMT1A)
Prukop T, et al.
Testing, 14(1), e0209752-e0209752 (2019)
Caenorhabditis elegans show preference for stimulants and potential as a model organism for medications screening
Engleman EA, et al.
Frontiers in Physiology, 9, 7-16 (2018)
Simon N Katner et al.
Behavioural brain research, 365, 7-16 (2019-02-26)
Alcohol use disorders (AUDs) cause serious problems in society and few effective treatments are available. Caenorhabditis elegans (C. elegans) is an excellent invertebrate model to study the neurobiological basis of human behavior with a conserved, fully tractable genome, and a
P Bienkowski et al.
European journal of pharmacology, 374(3), 321-327 (1999-07-28)
It has been repeatedly reported that endogenous opioid pathways play an important role in ethanol drinking behaviour. In line with these findings, a non-selective opioid receptor antagonist, naltrexone, seems to reduce relapse rates in detoxified alcoholics. The aim of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service