Skip to Content
Merck
All Photos(1)

Documents

N2538

Sigma-Aldrich

NS8593 hydrochloride

≥98% (HPLC), powder

Synonym(s):

N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-Benzimidazol-2-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3 · HCl
CAS Number:
Molecular Weight:
299.80
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +54 to +62°, c = 0.5 in methanol

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cl[H].C1C[C@@H](Nc2nc3ccccc3[nH]2)c4ccccc4C1

InChI

1S/C17H17N3.ClH/c1-2-8-13-12(6-1)7-5-11-14(13)18-17-19-15-9-3-4-10-16(15)20-17;/h1-4,6,8-10,14H,5,7,11H2,(H2,18,19,20);1H/t14-;/m1./s1

InChI key

VWEKCDTXUUPBNA-PFEQFJNWSA-N

Application

NS8593 hydrochloride has been used to study its effect on the growth of MDA-MB-231 cells (M.D. Anderson and metastasis breast cancer cells).

Biochem/physiol Actions

The compound (R)-N-(benzimidazol-2-yl)-1,2,3,4-tetrahydro-1-naphtylamine (NS8593) reversibly inhibited recombinant SK3-mediated currents (human and rat SK3) with potencies around 100 nM. NS8593 did not inhibit 125I-apamin binding. NS8593 decreased the calcium sensitivity by shifting the activation curve for Ca2+ to the right, only slightly affecting the maximal Ca2+-activated SK current. NS8593 inhibited all the SK1-3 subtypes Ca2+-dependently (Kd = 0.42, 0.60, and 0.73 μM, respectively, at 0.5 μM Ca2+. NS8593 did not affect the calcium-activated potassium channels of intermediate and large conductance (hIK and hBK channels, respectively). NS8593-mediated inhibition was prevented in the presence of a high concentration of the positive modulator NS309.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knockdown of the small conductance Ca2+-activated K+ channels is potently cytotoxic in breast cancer cell lines
Abdulkareem ZA, et al.
British Journal of Pharmacology, 173(1), 177-190 (2016)
Michiko Tashiro et al.
The journal of physiological sciences : JPS, 69(1), 97-102 (2018-06-18)
To examine whether TRPM7, a member of the melastatin family of transient receptor potential channels, is a physiological pathway for Mg2+ entry in mammalian cells, we studied the effect of TRPM7 regulators on cytoplasmic free Mg2+ concentration ([Mg2+]i) of rat
Alayna Mellott et al.
Pflugers Archiv : European journal of physiology, 472(11), 1589-1606 (2020-09-24)
TRPM7 is a cation channel-protein kinase highly expressed in T lymphocytes and other immune cells. It has been proposed to constitute a cellular entry pathway for Mg2+ and divalent metal cations such as Ca2+, Zn2+, Cd2+, Mn2+, and Ni2+. TRPM7
Francesca Luongo et al.
Nutrients, 10(6) (2018-06-19)
Intestinal magnesium (Mg) uptake is essential for systemic Mg homeostasis. Colon cells express the two highly homologous transient receptor potential melastatin type (TRPM) 6 and 7 Mg2+ channels, but their precise function and the consequences of their mutual interaction are
Goli Ardestani et al.
Cell calcium, 87, 102181-102181 (2020-02-26)
Prior to maturation, mouse oocytes are arrested at the germinal vesicle (GV) stage during which they experience constitutive calcium (Ca2+) influx and spontaneous Ca2+ oscillations. The oscillations cease during maturation but Ca2+ influx continues, as the oocytes' internal stores attain

Articles

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service