Skip to Content
Merck
All Photos(1)

Key Documents

N2019

Sigma-Aldrich

Nogalamycin

≥95%, from Streptomyces nogalater

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H49NO16
CAS Number:
Molecular Weight:
787.80
MDL number:
UNSPSC Code:
51111755
PubChem Substance ID:

biological source

Streptomyces nogalater

Assay

≥95%

form

solid

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

COC1C(C)OC(OC2CC(C)(O)C(C(=O)OC)c3cc4C(=O)c5c6OC7OC(C)(C(O)C(C7O)N(C)C)c6cc(O)c5C(=O)c4c(O)c23)C(OC)C1(C)OC

InChI

1S/C39H49NO16/c1-14-32(49-7)39(4,52-10)33(50-8)36(53-14)54-19-13-37(2,48)24(34(47)51-9)15-11-16-21(27(43)20(15)19)28(44)22-18(41)12-17-30(23(22)26(16)42)55-35-29(45)25(40(5)6)31(46)38(17,3)56-35/h11-12,14,19,24-25,29,31-33,35-36,41,43,45-46,48H,13H2,1-10H3

InChI key

KGTDRFCXGRULNK-UHFFFAOYSA-N

Biochem/physiol Actions

Anthracyclic antitumor antibiotic.
Anthracyclic antitumor antibiotic. It is used as a sequence specific DNA binding (intercalation) reagent, binds d(CGTACG), d(ATGCAT).

Other Notes

50mg
Keep container tightly closed in a dry and well-ventilated place.Light sensitive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thadee Grocholski et al.
Biochemistry, 49(5), 934-944 (2010-01-08)
SnoaB is a cofactor-independent monooxygenase that catalyzes the conversion of 12-deoxynogalonic acid to nogalonic acid in the biosynthesis of the aromatic polyketide nogalamycin in Streptomyces nogalater. In vitro (18)O(2) experiments establish that the oxygen atom incorporated into the substrate is
Ruogu Peng et al.
Organic letters, 14(8), 1962-1965 (2012-04-07)
A novel reductive Heck cyclization approach was developed in order to construct a model DEF-benzoxocin ring system that is present in nogalamycin, menogaril, and related anthracycline antitumor antibiotics.
Hanna Koskiniemi et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 65(Pt 3), 256-259 (2009-03-04)
12-deoxy-nogalonic acid oxygenase (SnoaB) catalyzes the oxygenation of 12-deoxy-nogalonic acid at position 12 to yield nogalonic acid, which is one of the steps in the biosynthesis of the polyketide nogalamycin in Streptomyces nogalater. SnoaB belongs to a family of small
Karin M Keller et al.
Journal of mass spectrometry : JMS, 40(10), 1362-1371 (2005-10-13)
The charge state-dependent dissociation of various DNA duplexes and drug/duplex complexes has been investigated using collisionally activated dissociation (CAD) in a quadrupole ion trap mass spectrometer (QIT-MS). Several non-self-complementary 14-residue oligonucleotides were employed, in addition to an array of known
Azmiri Sultana et al.
Acta crystallographica. Section D, Biological crystallography, 60(Pt 6), 1118-1120 (2004-05-26)
Nogalonic acid methyl ester cyclase (SnoaL) catalyzes the last ring-closure step in the biosynthesis of the polyketide antibiotic nogalamycin. Crystals of a complex of SnoaL with the substrate nogalonic acid methyl ester have been obtained using PEG 4000 as precipitant.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service