Skip to Content
Merck
All Photos(1)

Key Documents

N0765

Sigma-Aldrich

Nicotinic acid

suitable for plant cell culture

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

form

powder

mol wt

Mw 123.11 g/mol

technique(s)

cell culture | plant: suitable

color

white to faint yellow

pH

3.4 (20 °C, 10 g/L)

mp

236-239 °C (lit.)

solubility

1 M NaOH: soluble 50 mg/mL, clear, colorless to very faintly yellow

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)
rat ... Chrnb2(54239)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Graeme Semple et al.
Journal of medicinal chemistry, 51(16), 5101-5108 (2008-07-31)
The discovery and profiling of 3-(1H-tetrazol-5-yl)-1,4,5,6-tetrahydro-cyclopentapyrazole (5a, MK-0354), a partial agonist of GPR109a, is described. Compound 5a retained the plasma free fatty acid lowering effects in mice associated with GPR109a agonism, but did not induce vasodilation at the maximum feasible
John R Guyton et al.
Journal of the American College of Cardiology, 62(17), 1580-1584 (2013-08-07)
This study sought to examine the relationship between niacin treatment, lipoproteins, and cardiovascular (CV) outcomes in this secondary analysis of the AIM-HIGH (Atherothrombosis Intervention in Metabolic Syndrome With Low HDL/High Triglycerides and Impact on Global Health Outcomes) trial. During a
Koon K Teo et al.
Stroke, 44(10), 2688-2693 (2013-07-25)
In Atherothrombosis Intervention in Metabolic Syndrome with low HDL/High Triglycerides: Impact on Global Health Outcomes (AIM-HIGH) trial, addition of extended-release niacin (ERN) to simvastatin in participants with established cardiovascular disease, low high-density lipoprotein cholesterol, and high triglycerides had no incremental
Paul M Lavigne et al.
Journal of the American College of Cardiology, 61(4), 440-446 (2012-12-26)
This study sought to assess the efficacy of niacin for reducing cardiovascular disease (CVD) events, as indicated by the aggregate body of clinical trial evidence including data from the recently published AIM-HIGH (Atherothrombosis Intervention in Metabolic Syndrome with Low HDL/High
Sorin Tunaru et al.
Nature medicine, 9(3), 352-355 (2003-02-04)
Nicotinic acid (niacin), a vitamin of the B complex, has been used for almost 50 years as a lipid-lowering drug. The pharmacological effect of nicotinic acid requires doses that are much higher than those provided by a normal diet. Its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service