Skip to Content
Merck
All Photos(1)

Key Documents

N0411

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt

vial of 1 mg

Synonym(s):

β-NADPH, 2′-NADPH, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

15 VIALS
$330.00

$330.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
15 VIALS
$330.00

About This Item

Empirical Formula (Hill Notation):
C21H26N7Na4O17P3
Molecular Weight:
833.35
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

$330.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

form

solid

Quality Level

packaging

vial of 1 mg

SMILES string

[Na+].[Na+].[Na+].[Na+].[P](=O)([O-])([O-])O[C@H]1[C@@H](O[C@@H]([C@H]1O)CO[P](=O)([O-])O[P](=O)([O-])OC[C@@H]4O[C@@H]([C@H]([C@H]4O)O)N5C=C(CC=C5)C(=O)N)[n]2c3ncnc(c3nc2)N

InChI

1S/C21H30N7O17P3.4Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;;;/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);;;;/q;4*+1/p-4/t10-,11+,13-,14+,15-,16+,20-,21+;;;;/m0..../s1

InChI key

WYWWVJHQDVCHKF-MPUNMZHWSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivo by the pentose phosphate pathway (PPP).
Electron donor; cofactor for many redox enzymes including nitric oxide synthetase

Preparation Note

Chemically reduced

Other Notes

Packaged based on NADPH content as determined by UV-Vis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Chen et al.
iScience, 23(9), 101533-101533 (2020-10-22)
Lanthanide-based, Förster resonance energy transfer (LRET) biosensors enabled sensitive, time-gated luminescence (TGL) imaging or multiwell plate analysis of protein-protein interactions (PPIs) in living cells. We prepared stable cell lines that expressed polypeptides composed of an alpha helical linker flanked by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service