MX405MS20
Mix-n-Stain™ CF™ 405M Antibody Labeling Kit (5-20μg)
Select a Size
$184.00
Estimated to ship onJune 02, 2025
Select a Size
About This Item
$184.00
Estimated to ship onJune 02, 2025
Recommended Products
form
liquid
storage condition
protect from light
fluorescence
λex 408 nm; λem 452 nm
storage temp.
−20°C
General description
Application
Features and Benefits
Components
Analysis Note
Legal Information
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service