Skip to Content
Merck
All Photos(1)

Documents

MTOX1003

Sigma-Aldrich

MRP2 Knockout Caco-2 Cells

Human male colorectal tissue, adenocarcinoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

MRP2 Knockout Caco-2 Cells, one vial

biological source

human male colorectal tissue (Source disease: adenocarcinoma)

form

liquid

technique(s)

drug transporter assay: suitable
permeability assay: suitable

OMIM accession no.

application(s)

ADME/TOX

storage temp.

−196°C

Gene Information

human ... ABCC2(1244)

General description

The C2BBe1 cells, a subclone of Caco-2 cells, correspond to ATCC CRL-2102. The MRP2 knockout C2BBe1 cells are adenocarcinoma, epithelial cells from a human caucasian male (aged 72 years) with functional knockout of the ABCC2 (MRP2) efflux transporter.

Features and Benefits

The Caco-2 subclone C2BBe1 cells are ideal for transporter analysis as they express multiple transporters, are human derived, and grow in a homogenous monolayer that forms tight junctions necessary for efflux ratio analysis. Other benefits include:
  • A functional knockout of the MRP2 gene eliminates the reliance on chemical inhibitors to determine if a compound is an MRP2 substrate
  • The vial format enables the MRP2 knockout cells to be included in standard drug transporter protocols
  • Human assay with no interference from animal inhibitors
  • Overcome the limitations of RNAi and knockdown cell lines that arise from remaining transporter functionality

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • MTOX1003MRP2 Knockout Caco-2 Cells, one vial 1 vialSDS

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Artursson
Journal of pharmaceutical sciences, 79(6), 476-482 (1990-06-01)
A human intestinal cell line, Caco-2, was used as a model to study the passive diffusion of drugs across intestinal epithelium. The cells formed continuous monolayers when grown on permeable filters of polycarbonate. After 10 days in culture, the monolayers
S Yee
Pharmaceutical research, 14(6), 763-766 (1997-06-01)
To evaluate and optimize the use of Caco-2 cell monolayers to predict the in vivo absorption of a broad range of compounds in man. Caco-2 cells are derived from human adenocarcinoma colon cells and spontaneously differentiate when grown on porous
V Pade et al.
Journal of pharmaceutical sciences, 87(12), 1604-1607 (1999-04-03)
The objective of this investigation was to establish a relationship between drug permeability and solubility in vitro and the extent of drug absorption in humans. We selected drugs with varying permeabilities and solubilities with the aim of establishing a relationship
X Wu et al.
Pharmaceutical research, 17(2), 209-215 (2000-04-06)
The purpose of this study was to elucidate the mechanisms by which an HMG-CoA reductase inhibitor, atorvastatin (an organic acid with a pKa of 4.46), was transported in the secretory and absorptive directions across Caco-2 cell monolayers. Caco-2 cells were
M J Briske-Anderson et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 214(3), 248-257 (1997-03-01)
The Caco-2 cell line is used by many investigators as a model of the intestinal epithelium to study nutrient uptake and transport. Our goal was to create an awareness of inherent variabilities in the Caco-2 cell line which may influence

Articles

We presents an article on The Role of Intestinal Efflux Transporters In Drug Absorption.

Application note on Drug transport assays in a 96-well system using Millicell-96 System from Millipore.

Utilize these Caco-2 cell based assay tools for screening small molecule drug compounds prior to clinical studies and submission to regulatory agencies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service