Skip to Content
Merck
All Photos(1)

Key Documents

MSST0060

Sigma-Aldrich

SILuLite CST3, Cystatin C human

recombinant, expressed in E. coli, MS Protein Standard

Synonym(s):

CST3, Cystatin C

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$965.00

$965.00


Estimated to ship onApril 08, 2025



Select a Size

Change View
50 μG
$965.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.32

$965.00


Estimated to ship onApril 08, 2025


recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CST3(1471)

General description

SILuLite CST3 is a recombinant human protein expressed in E. coli. It consists of 120 amino acids, with a calculated molecular mass of 13.5 kDa. SILuLite CST3 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

Cystatin C is an inhibitor of cysteine proteases including cathepsin B (which has been identified as the most important ?-amyloid-degrading enzyme. Measurement of cystatin C in serum is replacing creatinine as an indicator of kidney function (glomerular filtration rate, GFR).

Sequence

SSPGKPPRLVGGPMDASVEEEGVRRALDFAVGEYNKASNDMYHSRALQVVRARKQIVAGVNYFLDVELGRTTCTKTQPNLDNCPFHDQPHLKRKAFCSFQIYAVPWQGTMTLSKSTCQDA

Physical form

Supplied as a lyophilized powder containing tris buffered saline and methionine.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service