Skip to Content
Merck
All Photos(1)

Key Documents

MSST0043

Sigma-Aldrich

SILuProt TIMP1, Metalloproteinase inhibitor 1 human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$903.00

$903.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$903.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.54

$903.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

General description

SILuProt TIMP1 is a recombinant, stable isotope-labeled human TIMP1 which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of TIMP1 in mass-spectrometry. SILuProt TIMP1 is a protein of 184 amino acids , with a calculated molecular mass of 20.7 kDa.

Biochem/physiol Actions

TIMP-1 is glycoprotein that is over-expressed in the supernatant of tissue extracts of breast, gastric, colorectal, and hepatocellular carcinomas. It belongs to the family of “tissue inhibitors of metalloproteinases,” a group of proteins that help regulate bone turnover. TIMP-1 serum levels are significantly associated with HER2 extracellular domain (ECD)-positivity and poorer disease-free survival among primary breast cancer patients with HER2 overexpression. High levels of serum TIMP-1 correlate with advanced disease and predict for poor survival in patients with multiple myeloma treated with bortezomib and/or IMiDs during their disease course.

Sequence

CTCVPPHPQTAFCNSDLVIRAKFVGTPEVNQTTLYQRYEIKMTKMYKGFQALGDAADIRFVYTPAMESVCGYFHRSHNRSEEFLIAGKLQDGLLHITTCSFVAPWNSLSLAQRRGFTKTYTVGCEECTVFPCLSIPCKLQSGTHCLWTDQLLQGSEKGFQSRHLACLPREPGLCTWQSLRSQIA

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service