Skip to Content
Merck
All Photos(1)

Key Documents

MSP3L

Sigma-Aldrich

MS PhosphoMix 3 Light

Phosphopeptide Standard for MS

Synonym(s):

Light PhosphoMix, MS Phosphorylation Mix, PhosphoMix 3 Light

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$381.00
5 X 1 VIAL
$1,540.00

$381.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
$381.00
5 X 1 VIAL
$1,540.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

$381.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

quality

Phosphopeptide Standard for MS

Quality Level

analyte chemical class(es)

amino acids, peptides, proteins

packaging

pkg of 200 pmol total phosphopeptides

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

food and beverages

format

multi-component solution

storage temp.

−20°C

General description

The MS PhosphoMix line of products allows for the testing of the strengths and weaknesses of phosphopeptide sample processing, mass spectrometry analysis and instrument configurations. The mixes are produced from synthetic phosphopeptides with sequences derived from naturally occurring peptides as identified by Mann et al. in HeLa cells.[1] Because the sequences are derived from mammalian cells, many natural phosphorylation motifs, such as those that present an abundance of proline, are represented.[2] Additionally, the phosphopeptide distribution in each mix has been chosen to present a broad range of characteristics, including ionizability, LC retention time, charge state, and isoelectric point. Finally, PhosphoMix-1, 2, and 3 were designed in a complementary fashion, as highlighted on the following page. For example, all three mixes contain peptides of the same sequence with different sites of phosphorylation.

Each of the three phosphopeptides mixes are available in their naturally occurring isotopic abundances (light) or as stable isotope enriched versions (heavy), making the set of products highly amenable to quantitative analyses, allowing users to compare recovery between workflows or techniques.

  • Naturally occurring peptide sequences
  • Broad range of peptide characteristics
  • Complementary product designs
  • Available in light and heavy versions

More info and FASTA file

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorte B Bekker-Jensen et al.
Nature communications, 11(1), 787-787 (2020-02-09)
Quantitative phosphoproteomics has transformed investigations of cell signaling, but it remains challenging to scale the technology for high-throughput analyses. Here we report a rapid and reproducible approach to analyze hundreds of phosphoproteomes using data-independent acquisition (DIA) with an accurate site
Jordy J Hsiao et al.
Analytical chemistry, 90(15), 9457-9464 (2018-07-07)
Phosphorylated compounds and organic acids with multiple carboxylate groups are commonly observed to have poor peak shapes and signal in LC/MS experiments. The poor peak shape is caused by the presence of trace metals, particularly iron, contributed from a variety
Evgeny Kanshin et al.
Journal of proteome research, 12(6), 2905-2913 (2013-04-24)
Phosphorylation is a reversible protein modification that regulates major cellular processes such as cell division, growth, and differentiation through highly dynamic and complex signaling pathways. Large-scale phosphoproteomics analyses have been greatly facilitated using affinity chromatography such as metal oxide affinity
Alexander R Ivanov et al.
Proteomics, 13(6), 904-909 (2013-01-16)
Proteomics is a rapidly transforming interdisciplinary field of research that embraces a diverse set of analytical approaches to tackle problems in fundamental and applied biology. This viewpoint article highlights the benefits of interlaboratory studies and standardization initiatives to enable investigators
Fernando Martínez-Montañés et al.
Cell reports, 32(6), 108024-108024 (2020-08-14)
The ability to remodel lipid metabolism under changing conditions is pivotal for cellular functionality and homeostasis. Here, we characterize the regulatory landscape of phosphorylation-based signaling events across the life cycle of Saccharomyces cerevisiae and determine its impact on the regulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service