Skip to Content
Merck

Skip To

MSP09

Membrane Scaffold Protein 1E3D1 D73C

recombinant, expressed in E. coli, Cysteine substituted at position 73

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mg
Check Cart for Availability
$693.00

About This Item

NACRES:
NA.26
UNSPSC Code:
12352200
Form:
buffered aqueous solution
Assay:
≥90% (SDS-GE)
Recombinant:
expressed in E. coli

$693.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

assay

≥90% (SDS-GE)

form

buffered aqueous solution

shipped in

ambient

storage temp.

−20°C

General description

Nanodisc technology is an approach to render membrane proteins soluble in aqueous solutions in a native-like bilayer environment, where the membrane proteins remain stable and active. The Nanodisc concept is derived from high density lipoprotein (HDL) particles and their primary protein component, apolipoprotein. The Nanodisc is a non-covalent structure of phospholipid bilayer and membrane scaffold protein (MSP), a genetically engineered protein, which mimics the function of Apolipoprotein A-1 (ApoA-1).

The first MSP, MSP1, was engineered with its sequence based on the sequence of A-1, but without the globular N-terminal domain of native A-1. The MSP1E3D1 D73C variant of MSP1 differs from MSP1 in the following facets:
  • It deletes the first 11 amino acids in the Helix 1 portion (referred to as “H0.5” in the accompanying figure) of the original MSP1 sequence3 (which is known separately as MSP1D1).
  • It repeats the Helix 4 (H4), Helix 5 (H5) and Helix 6 (H6) sequences of the original MSP1 sequence between the parent Helix 6 (H6) and Helix 7 (H7) segments of MSP1D1.
  • It substitutes a cysteine (C) residue for an aspartic acid (D) residue in the Helix 2 (H2) portion of the protein, at position 73 of the original native A-1 sequence.
  • The initial histidine-tag is one amino acid shorter.

Application

For guidelines on the use of this and other MSP′s to prepare Nanodiscs, please visit our Protocols for Membrane Scaffold Proteins and Nanodisc Formation page.

Legal Information

Nanodisc technology, and many of its uses, are covered by the following patents held by the University of Illinois.
  • 7,691,414 Membrane scaffold proteins
  • 7,662,410 Membrane scaffold proteins and embedded membrane proteins
  • 7,622,437 Tissue factor compositions and methods
  • 7,592,008 Membrane scaffold proteins
  • 7,575,763 Membrane scaffold proteins and tethered membrane proteins
  • 7,083,958 Membrane scaffold proteins
  • 7,048,949 Membrane scaffold proteins


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSP05MSP03MSP10
recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

assay

≥90% (SDS-GE)

assay

≥90% (SDS-GE)

assay

≥90% (SDS-GE)

assay

≥90% (SDS-GE)

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

Nanodisc technology aids membrane protein solubilization, overcoming associated challenges in diverse protein classes.

Related Content






Global Trade Item Number

SKUGTIN
MSP09-5MG04061831159691

Questions

Reviews

No rating value

Active Filters