Skip to Content
Merck
All Photos(1)

Key Documents

MSANGERV

Sigma-Aldrich

Sanger Arrayed Whole Genome Lentiviral CRISPR Library

Mouse, Virus Format

Synonym(s):

Arrayed CRISPR library, Sanger CRISPR library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105904

Quality Level

packaging

pkg of 10 μL (384-well plate)

concentration

1x106  VP/ml (via p24 assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Two leaders in genome editing, Sigma-Aldrich and the Wellcome Trust Sanger Institute, have joined forces to make the first ever arrayed lentiviral CRISPR knockout libraries. Based upon validated techniques published in the literature, the Sanger CRISPR libraries will put your lab at the forefront of the race to make the next big discovery.

Application

Functional Genomics/Screening /Target Validation

Features and Benefits

  • Vector: U6-gRNA/PGK-Puro-2A-BFP (gRNA only)
  • Simplify the workflow with puromycin selection
  • Illuminate CRISPR-expressing cells with BFP

Additional Features
  • Better, not bigger: Two optimized clones per mouse gene reduces the time, cost, and scale of screening experiments
  • Ready-to-screen: Clones are arrayed in a robotics-friendly 384-well format for high throughput screening
  • Collaborative: Real-time, library validation continues

For detailed information on the Sanger library, click here

Packaging

384 well plates

Components

Lentivirus Transduction Particles of gRNA-only lenti-based vectors. 2 knockout clones for every human protein-coding gene. Nearly 40,000 sequence confirmed clones


Request a Quote or More Information

Physical form

10μl of lentivirus at ≥106 particles/ml in ~120×384 well plates

Other Notes

This product is for R&D use only, not for drug, household, or other uses. Lentivirus manufacturing is achieved by using 2nd generation packaging plasmids on a 3rd generation lenti-based vector. Though the lentiviral transduction particles produced are replication incompetent, it is recommended that they be treated as Risk Group Level 2 (RGL-2) organisms in laboratory handling. Follow all published RGL-2 guidelines for laboratory handling and waste decontamination.

Recommended products

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing the genome editing toolbox: genome wide CRISPR arrayed libraries.
Metzakopian, E. et al
Scientific Reports, 7 (2017)

Articles

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Get tips for handling lentiviruses, optimizing experiment setup, titering lentivirus particles, and selecting helpful products for transduction.

Protocols

Learn about Sanger Sequencing steps or the chain termination method and how DNA sequencing works and how to read Sanger Sequencing results accurately for your research.

FACS sorts cells based on light scattering and fluorescence for objective cell analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service