The method of inactivation and the specific formulation are considered proprietary for this product.
MBD0021
Inactivated Bacillus subtilis
Suitable for PCR, sequencing and NGS, >10^8 bacteria/ml
About This Item
Recommended Products
Quality Level
form
powder
concentration
>10^8 bacteria/ml
technique(s)
DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Read here how to use our standards to ensure data integrity for your microbiome research.
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Turn to our Next-Gen Sequencing Oligos (NGSO) as a potential solution if you are experiencing high levels of adapter dimer formation with custom adapters sourced from other vendors.
DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.
-
Hi. What is the method of inactivation? are there any detergents in the preparation? Thanks.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service