Skip to Content
Merck
All Photos(1)

Key Documents

MBD0019

Sigma-Aldrich

Microbial DNA standard from Burkholderia pyrrocinia

Suitable for PCR, sequencing and NGS, 10 ng/μL

Sign Into View Organizational & Contract Pricing

Select a Size

0.3 μG
$335.00

$335.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
0.3 μG
$335.00

About This Item

UNSPSC Code:
41105500
NACRES:
NA.24

$335.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

form

liquid

concentration

10 ng/μL

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses. Burkholderia pyrrocinia genomic DNA microbial standard can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.

Burkholderia cepacia complex (Bcc) species are a group of Gram-negative opportunistic pathogens that chronically infect the airways of cystic fibrosis patients, but they can also infect patients with various types of immunosuppressive disorders. Bcc members are multidrug resistant bacteria that have the ability to persist in the infected host and also elicit robust inflammatory responses. It currently includes 20 closely related, but distinct, species of bacteria, with a wide environmental distribution.

B. cepacia has an unusually large genetic make up with a total DNA content greater than 7 megabases and as many as three megabase-sized replicons. This large genetic make up accounts for its microbiological versatility. Advances in the taxonomy of Bcc revealed that it comprises at least nine genetically closely related species, including B. pyrrocinia, which can be differentiated on the basis of molecular and biochemical tests.

Read here how to use our standards to ensure data integrity for your microbiome research.

Application

The genomic DNA is provided at >=10 ng/μL concentration in TE buffer pH 8.0. It is recommended to avoid freeze thaw cycles of this product.
Suitable for Quantitative standard for PCR, Sequencing and NGS

Features and Benefits

  • Individual microbial standard for microbiomics and meta-genomics workflow
  • Suitable standard for PCR, sequencing and NGS
  • Improve Bioinformatics analyses
  • Increases reproducibility
  • Compare results lab to lab

Physical form

Liquid - The genomic DNA is provided at >=10 ng/μL concentration in TE buffer pH 8.0

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service