MAK554
Fluorimetric Acetylcholinesterase Assay Kit
sufficient for 200 fluorometric tests
About This Item
Recommended Products
detection method
fluorometric
relevant disease(s)
hematological disorder; neurological disorders
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Principle
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service