Skip to Content
Merck
All Photos(1)

Key Documents

MAK551

Sigma-Aldrich

Butyrylcholinesterase (BChE) Activity Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104
NACRES:
NA.21
Pricing and availability is not currently available.

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; obesity; cardiovascular diseases

shipped in

dry ice

storage temp.

−20°C

General description

Butyrylcholinesterase (EC 3.1.1.8; BChE), also known as pseudocholinesterase or plasma cholinesterase, is mainly synthesized in liver and present in blood. BChE is a nonspecific cholinesterase enzyme and can hydrolyze many different choline esters, serving as the first line of defense against toxic compounds reaching the bloodstream. ​

The Butyrylcholinesterase Activity Assay Kit is based on a synthetic butyrylthiocholine based substrate, which can be hydrolyzed by BChE and produce thiocholine. Thiocholine can react with 5, 5′-dithiobis (2-nitrobenzoic acid) (DTNB) and generates a yellow chromophore that can be detected at 410 nm. The assay is convenient, sensitive and can detect as low as 6 mU/mL in a variety of samples.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service