MAK488
Phosphate Assay Kit
sufficient for 500 colorimetric tests
Synonym(s):
Phosphate Colorimetric Test Kit
About This Item
Recommended Products
Quality Level
application(s)
agriculture
cosmetics
environmental
food and beverages
pharmaceutical
detection method
colorimetric
shipped in
ambient
storage temp.
2-8°C
Related Categories
General description
Features and Benefits
- Linear detection range: 0.3 µM (0.0028 mg/dL) to 50 µM (0.47 mg/dL) phosphate.
- Compatible with high-throughput handling systems.
- The optimized formulation has greatly enhanced reagent and signal stability.
- Low interference in biological samples. No pretreatments are needed.
Suitability
Principle
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service