MAK431
Methionine Adenosyltransferase Activity Assay Kit (Colorimetric)
sufficient for 100 colorimetric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
application(s)
pharmaceutical
detection method
colorimetric
relevant disease(s)
genitourinary disorders; cancer
storage temp.
−20°C
General description
Methionine Adenosyltransferases (MATs), also known as Adenosylmethionine Synthetases are a family of enzymes that synthesize S-adenosyl-L-methionine (SAMe) from L-methionine and ATP. SAMe is a primary biochemical alkylation agent (one of only two possible methyl group donors) and is also a vital metabolic precursor of the trans-sulfuration and polyamine synthesis (aminopropylation) pathways. All organisms express at least one MAT enzyme. In mammals, three isozymes of MAT have been identified. MAT1 and MAT3 isozymes are predominantly expressed in the liver, whereas MAT2A is expressed in most tissues. Upregulation of the MAT2A isozyme has been linked to several human diseases. MAT2A has become a popular drug target for novel cancer therapeutics as well as for hepatic fibrosis and non-alcoholic fatty liver disease.
Features and Benefits
Methionine Adenosyltransferase Activity Assay Kit:
- Compatible with high-throughput handling systems.
- Limit of quantification of 2 mU MAT activity per well.
- Homogeneous assay.
- Does not require complicated sample processing.
Suitability
The kit is suitable for the rapid assessment of MAT activity in human or animal soft tissue (i.e. brain, liver, lung, etc.) homogenates and cultured cell lysates (adherent or suspension cells) or heterologously expressed recombinant MAT preparations.
Principle
The Methionine Adenosyltransferase Activity Assay Kit enables the rapid measurement of MAT activity in complex biological matrices. The assay is based on the detection of pyrophosphate, which is generated stoichiometrically during the generation of SAMe. Pyrophosphate is enzymatically metabolized to an intermediate product, which reacts with the probe to form a stable chromophore that is detected by absorbance at A570 nm.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service