Skip to Content
Merck
All Photos(1)

Key Documents

MAK418

Sigma-Aldrich

AMP Colorimetric Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

detection method

colorimetric

relevant disease(s)

neurological disorders; aging/geriatric diseases; cancer

storage temp.

−20°C

General description

Adenosine Monophosphate (AMP), also known as 5′-adenylic acid, consists of adenine, ribose and phosphate. AMP can be produced during ATP synthesis by the enzyme adenylate kinase by combining two ADP molecules, as a result of the hydrolysis of ATP/ADP, or when RNA is broken down. AMP plays an important role in many cellular metabolic processes, such as Ca2+ signaling, cell migration, cytokine secretion etc. AMP is an activator of AMP-activated protein kinase, which regulates glucose uptake, fatty acid uptake, fatty acid oxidation etc. AMP levels can be measured by luciferase/luciferin mediated assays. However, luciferase signal is unstable and luminescent equipment is generally not available in most laboratories.

Features and Benefits

AMP Assay Kit:
  • Compatible with high-throughput handling systems.
  • Sensitive, detecting AMP levels lower than 10 μM in various sample types.

Suitability

The kit is suitable for the measurement of AMP levels in various cell (adherent or suspension cells: e.g. HeLa, HEK239, Jurkat cells) and tissue lysates (e.g. liver, kidney, etc.), the study of the regulation of AMPK by AMP and for the mechanistic study of key cellular processes such as Ca2+ signaling, glucose uptake, lipid uptake, etc.

Principle

In this assay, AMP is converted to pyruvate in the presence of pyrophosphate and phosphoenolpyruvate. This is followed by a set of enzymatic reactions to generate a colored product with a strong absorbance at A570 nm. The absorbance is proportional to the amount of AMP present in samples.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service