Skip to Content
Merck
All Photos(1)

Key Documents

MAK404

Sigma-Aldrich

Carbonic Anhydrase (CA) Activity Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Synonym(s):

CA Enzyme Activity Assay Kit (Colorimetric)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; cancer; hematological disorder

storage temp.

−20°C

General description

Carbonic anhydrases (CA) are zinc enzymes present in both prokaryotes and eukaryotes. They catalyze efficiently the reversible hydration of CO2 to bicarbonate. Their important patho-physiological roles in respiration, pH and CO2 homeostasis, secretion, gluconeogenesis, ureagenesis etc. make it an important drug target. Characteristic increase and decrease of CA Activity is observed in different physiological conditions and diseases such as anemia, Thalassemia, Hypothyroidism, Hyperthyroidism, and in several cases of lung and liver diseases and Leukemia.

Features and Benefits

Carbonic Anhydrase (CA) Activity Assay Kit:
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the detection of carbonic anhydrase activity in hemolysates and serum, as well as the determination of enzymatic activity of purified CA.

Principle

The assay utilizes the esterase activity of an active CA on an ester substrate which releases a chromophore.The released product can be easily quantified using a spectrophotometric multi-well plate reader. In the presence of a CA specific inhibitor, the enzyme loses its activity which results in a decrease of absorbance.To measure specific CA activity, the CA inhibitor Acetazolamide is included.Acetazolamide is a potent inhibitor of CA I, CA II, CA IV, CA IX, CA XII etc. except CA III.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is MAK404-1KT Carbonic Anhydrase Activity Assay Kit (Colorimetric) suitable for use with bacteria and mammalian tissue lysates?

    1 answer
    1. The Carbonic Anhydrase (CA) Activity Assay Kit (Colorimetric) has not been tested with bacteria samples, so there are no specific recommendations available for this application. Additionally, this kit has not been tested with mammalian lysates. The lysis buffer required for the assay may be incompatible with the buffers provided, and the substrate may not be compatible or may cross-react with other biological components found in a lysate.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service