Skip to Content
Merck
All Photos(1)

Key Documents

MAK326

Sigma-Aldrich

Indole Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$250.00

$250.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$250.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$250.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases

storage temp.

2-8°C

General description

Indole is the primary product of tryptophan breakdown by tryptophanase. Structurally, it is characterized by a benzene ring fused to a pyrrole ring, containing a five-membered nitrogen-containing ring. This simple structure forms the core of a vast array of natural and synthetic compounds with a wide range of biological activities. In bacteria, indole is a critical signaling molecule, produced as a metabolite during the breakdown of the amino acid tryptophan. It plays a pivotal role in bacterial behavior, influencing biofilm formation, spore germination, and the expression of virulence factors. In the context of metabolism in larger organisms, indole derivatives are integral to the functioning of the central nervous system, serving as precursors for the synthesis of neurotransmitters such as serotonin and melatonin. These compounds are consequential in regulating mood, appetite, sleep, and circadian rhythms. Indole metabolism in the gut microbiota has been implicated in maintaining the integrity of the intestinal epithelial barrier and modulating the immune response, reflecting the complexity and importance of this molecule in biological systems.

Application

The Nitric Oxide Assay Kit may be used for:
  • Gastrointestinal Disease Research
  • Bacterial Culture Analysis

Features and Benefits

Quick and Sensitive: Detect Indole levels ranging from 3 - 100 μM with a short 10 minute procedure, providing both convenience and sensitivity.

Simplified Process: Enjoy a hassle-free procedure that requires the addition of a single working reagent, saving you time and effort.

Suitability

Suitable for indole determination in biological samples (e.g. indole produced by indole positive bacteria).

Principle

The indole assay kit is based on a modified version of Ehlrich′s and Kovac′s reagents, which reacts with indole to produce a colored compound at 565 nm. The intensity of this colored compound is directly proportional to the indole in the sample. Linear detection range from 3 to 100 μM indole in 96-well plate assay.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–6 of 6 Questions  
  1. If the test assumes it's indoxyl sulfate, a high conc. say 80-90 um would indicate toxicity, considering its a uremic toxin? Higher ILA/IPA on other hand would mean a beneficial indole.

    1 answer
    1. This kit is not intended to categorize a test sample as toxic. The test is designed to measure urinary Indican (Indoxyl sulfate) and assumes that most of the Indole in the sample exists as urinary Indican (Indoxyl sulfate). The test does not distinguish between Indole, Indican, or urinary Indican (Indoxyl sulfate), so it will likely detect other species such as Indolepyruvic acid, Indolelactic acid, and Indolepropionic acid if the reactive carbon is available. The test does not convert the indole derivative before the DMAB reaction.

      Helpful?

  2. Is any up front extraction of the biological sample necessary before running the assay (example: solvent extraction)?

    1 answer
    1. The necessity for extraction often depends on the type of biological sample (e.g., serum, plasma, tissue, or cell lysates). Solid tissues may require extraction, while fluid samples may not.

      Helpful?

  3. Is this working in 560nm in microplate reader?

    1 answer
    1. For optimal results, the plate should be read at 565 nm, as per the kit protocol.

      Helpful?

  4. Will this kit react with tryptophan alone. For example will it react in the presence of 100µM Trp?

    1 answer
    1. This kit will detect and quantify indole only. Please consider alternate product MAK523 if the interest is in assaying tryptophan concentration. Please see the link below to review this product:
      https://www.sigmaaldrich.com/product/sigma/mak523

      Helpful?

  5. Does this kit only measure indole, or does it detect a wide variety of indole-containing compounds such as indolepyruvic acid, indolelactic acid, and indolepropionic acid?

    1 answer
    1. The kit has not been tested with those compounds. The test is specifically designed to measure urinary indican (indoxyl sulfate), assuming that most of the indole in the sample exists as urinary indican (indoxyl sulfate).

      Helpful?

  6. Does this assay detect both indole and all its derivatives? Or is there a specificity towards some indoles?

    1 answer
    1. Unfortunately, this kit has not been tested with Indole compounds such as indolepyruvic acid, indolelactic acid, and indolepropionic acid. The test is designed to measure urinary indican (indoxyl sulfate). It assumes that most of the indole in the sample exists as urinary indican (indoxyl sulfate). The test does not distinguish between indole, indican, or urinary indican (indoxyl sulfate) so it will likely pick up the species enumerated if the reactive carbon is available. The test does not convert the indole derivative before the DMAB reaction.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service