Skip to Content
Merck
All Photos(1)

Key Documents

MAK318

Sigma-Aldrich

α−Mannosidase Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$580.00

$580.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$580.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$580.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

detection method

colorimetric

relevant disease(s)

neurological disorders

storage temp.

2-8°C

General description

α-Mannosidase (AMA) is an enzyme which catalyzes the cleavage of the alpha form of mannose. α-Mannosidase assists in the breakdown of complex sugars from glycoproteins in the lysosome. Defective AMA or deficient AMA activity causes α-mannosidosis and leads to deterioration of the central nervous system in children.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for determining α-Mannosidase (AMA) activity in a variety of biological samples such as plasma, serum, tissue, and cell culture media.

Principle

The α-Mannosidase Activity Assay Kit provides a simple and high-throughput adaptable assay for determining α-Mannosidase activity in a variety of biological samples such as plasma, serum, tissue, and cell culture media. In this assay 4-nitrophenol is cleaved from a synthetic substrate. Nitrophenol becomes intensely colored with the addition of the stop reagent, producing a colorimetric (405 nm) result, directly proportional to the enzyme activity in the sample.
One unit of AMA is the amount of enzyme that will convert 1.0 μmole of 4-Nitrophenyl-α-D-mannopyranoside to 4-Nitrophenol and α-D-Mannose per minute at 25 °C and pH 4.5. The kit has a linear detection range of 1–250 unit/L for a 10 minute reaction in a 96 well format.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service