Skip to Content
Merck
All Photos(1)

Key Documents

MAK317

Sigma-Aldrich

Sorbitol Dehydrogenase Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$559.00

$559.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$559.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$559.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; gastrointestinal diseases

storage temp.

−20°C

General description

Sorbitol Dehydrogenase (SDH) is an enzyme that catalyzes the interconversion of sorbitol and fructose. Elevated blood serum SDH levels indicate liver damage. Thus, SDH plays an important role in the diagnosis of liver disease, especially in combination with aminotransferases. SDH levels are also measured to evaluate diabetic complications such as proliferative diabetic retinopathy.[1][2]

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for determining sorbitol dehydrogenase (SDH) activity in a number of biological samples such as plasma, serum, urine, tissue, and culture media.

Principle

Sorbitol dehydrogenase assay is a non-radioactive, colorimetric assay. It is based on the reduction of the tetrazolium salt MTT (3-[4, 5-dimethylthiazol-2-yl]-2, 5 diphenyl tetrazolium bromide) in a NADH-coupled enzymatic reaction. The reduced form of MTT exhibits an absorption maximum at 565 nm and the increase in absorbance at 565 nm is directly proportional to the enzyme activity. This assay is based on a kinetic reaction. Sorbitol dehydrogenase (SDH) activity can be determined in a number of biological samples (e.g., plasma, serum, urine, tissue, and culture media).

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The current state of serum biomarkers of hepatotoxicity.
Ozer J, et al.
Toxicology, 245(3), 194-205 (2008)
Pathophysiology of diabetic retinopathy.
Tarr J M, et al.
ISRN ophthalmology (2013)
Luciana F Brito et al.
Applied microbiology and biotechnology, 104(11), 5095-5106 (2020-04-11)
Gene repression using the endonucleolytically deactivated dCas9 protein and sgRNAs (CRISPR interference or CRISPRi) is a useful approach to study gene functions. Here, we established CRISPRi in Paenibacillus sonchi genomovar Riograndensis SBR5, a plant growth promoting bacterium. CRISPRi system with

Questions

  1. What are the recommended methods for homogenizing or lysing cells in a 96-well plate for the MAK317 Sorbitol Dehydrogenase assay kit? The manual advises against using proteolytic enzymes and suggests using a rubber policeman, which is not suitable for a 96-well plate. Are there any recommendations for a mild lysing buffer with minimal impact on the assay outcome? Additionally, is it possible and recommended to run the assay in the same 96-well plate, or is transferring the samples to a new plate necessary?

    1 answer
    1. The supplier advises that they have not yet validated any lysis buffers for use with cell samples in conjunction with this kit. Additionally, they recommend using an untreated, clear, flat-bottom plate for colorimetric measurements. Therefore, it would be preferable to transfer the samples to such a plate for running the assay.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service