Skip to Content
Merck
All Photos(1)

Key Documents

MAK306

Sigma-Aldrich

LCFC-LCAT Acyltransferase Activity Assay

Supplied by Roar Biomedical, Inc., sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,040.00

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 KIT
$1,040.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

usage

sufficient for 100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

cardiovascular diseases

storage temp.

2-8°C

General description

The plasma protein lecithin:cholesterol acyltransferase (LCAT) catalyzes the transfer of an acyl group from the sn2 position of phosphatidylcholine to the 3-hydroxyl group of cholesterol forming cholesteryl ester. This activity occurs on the surface of high density lipoprotein (HDL) and the cholesteryl esters formed by LCAT are incorporated into the core of HDL.

Features and Benefits

  • Non-enzymatic determination of cholesteryl ester formation.
  • Not affected by LCAT inhibitors or other chemical compounds.
  • Dilution or non-dilution type assay.
  • Assay components stable for up to one year.
  • Compatible with high-throughput handling systems.

Suitability

Suitable for high-throughput screening, mechanism of action studies and structureactivity relationship (SAR) work of LCAT in plasma and serum

Principle

The LCFC-LCAT Acyltransferase Activity Assay is a fluorometric assay useful for measuring the acyltransferase activity of LCAT in serum or plasma. The method detects changes in LCAT free cholesterol (LCFC) levels in the sample without the use of cholesterol oxidase, peroxidase, or the generation of hydrogen peroxide. Detection is not affected by iodoacetate or other LCAT inhibitors. LCAT activity results in a fluorometric (λEx = 320 nm/λEm = 405 nm) product proportional to the amount of free cholesterol present.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Liu et al.
PloS one, 12(1), e0169939-e0169939 (2017-01-21)
Hypertriglyceridemia is an independent risk factor for cardiovascular disease. Apolipoprotein C-II (APOC2) is an obligatory cofactor for lipoprotein lipase (LPL), the major enzyme catalyzing plasma triglyceride hydrolysis. We have created an apoc2 knockout zebrafish model, which mimics the familial chylomicronemia
Deficient cholesterol esterification in plasma of apoc2 knockout zebrafish and familial chylomicronemia patients.
Liu C, et al.
PLoS ONE, 12(1), e0169939-e0169939 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service