Skip to Content
Merck
All Photos(1)

Key Documents

MAK228

Sigma-Aldrich

Calpain Activity Fluorometric Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; neurological disorders; cancer; cardiovascular diseases

storage temp.

−70°C

General description

Activation of calpain is involved in many forms of physiological and pathological processes (e.g., apoptosis). Calpain activation requires cell membrane and Ca2+, and activated calpain is released into cytosol. The Calpain Activity Assay Kit provides optimized buffers and reagents for a convenient measurement of calpain activity.

Suitability

Suitable for analyzing calpain activity in apoptotic and other samples.

Principle

Calpain Activity Fluorometric Assay Kit is based on the cleavage of the calpain substrate, Ac-LLY-AFC. The Ac-LLY-AFC substrate emits blue light (max = 400 nm); upon cleavage by calpain, free AFC emits a yellow-green fluorescence ( max = 505 nm), which can be quantified using a fluorometer or a fluorescence plate reader. Comparison of the fluorescence intensity from a treated sample with a normal control allows determination of the changes in calpain activity.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judy Kim et al.
Bioscience reports, 40(1) (2020-01-09)
A key mechanism mediating cellular adaptive responses to hypoxia involves the activity of hypoxia-inducible factor 1 (HIF-1), a transcription factor composed of HIF-1α, and HIF-1β subunits. The classical mechanism of regulation of HIF-1 activity involves destabilisation of HIF-1α via oxygen-dependent
Huilin Cheng et al.
Food science of animal resources, 42(2), 266-279 (2022-03-22)
This study was conducted to evaluate the proteolysis trends and change in meat quality during 10 days of cold storage in duck M. pectoralis major (PM) and M. iliotibialis (IL). Duck IL had a higher pH and greater degree of
Alexander Otahal et al.
Scientific reports, 10(1), 959-959 (2020-01-24)
Hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase inhibitors (statins) have been shown to overcome tyrosine kinase inhibitor (TKI) resistance in epithelial growth factor receptor (EGFR) mutated non-small cell lung cancer (NSCLC) cells in vivo and in vitro. However, little is known about the
Huilin Cheng et al.
Poultry science, 101(12), 102185-102185 (2022-10-12)
The proteolysis trends and meat quality of the chicken pectoralis major (PM) and iliotibialis (IL) muscles stored at 4°C for 7 d were investigated. After 7 d of storage, the purge loss was higher (P < 0.05) in PM than

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service