Skip to Content
Merck
All Photos(2)

Key Documents

MAK208

Sigma-Aldrich

GPDH Activity Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

Glycerol 3-Phosphate Dehydrogenase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

detection method

colorimetric

storage temp.

−20°C

Gene Information

General description

Glycerol 3-phosphate dehydrogenase (GPDH; EC 1.1.1.8) catalyzes the reversible conversion of dihydroxyacetone phosphate and NADH to glycerol-3-phosphate and NAD+. It forms a glycerol phosphate shuttle and facilitates the transfer of reduced equivalents across the mitochondria to the cytosol. GPDH is important for both carbohydrate and lipid metabolism. Mutations in GPDH gene result in transient infantile hypertriglyceridemia.

Application

GPDH (glyceraldhyde-3-phoshate dehydrogenase) activity assay kit has been used to measure the serum and hepatic concentrations of GPDH in rabbit.

Suitability

Suitable for the detection of Glycerol-3-Phosphate Dehydrogenase Activity in tissue and cell culture samples.

Principle

The Glycerol 3-Phosphate Dehydrogenase Activity Colorimetric Assay Kit provides a simple and sensitive procedure for measuring GPDH activity in a variety of tissues and cells. GPDH activity is determined by measuring a colorimetric product with absorbance at 450 nm (A450) proportional to the enzymatic activity present. One unit of GPDH is the amount of enzyme required to generate 1.0 μmole of NADH per minute at pH 8 at 37 °C.

Kit Components Only

Product No.
Description

  • GPDH Assay Buffer

  • GPDH Substrate

  • GPDH Probe

  • NADH Standard

  • GPDH Positive Control

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of adenine nucleotide translocase and glycerol 3-phosphate dehydrogenase expression by thyroid hormones in different rat tissues.
DUMMLER K, et al.
The Biochemical Journal, 317(3), 913-918 (1996)
B. Elmahdi, MA Al-Omair, AA El-Bessoumy and SM El-Bahr.
Thomson I S I
International Journal of Pharmaceutics, 12(4), 429-434 (2016)
A compound heterozygous mutation in GPD1 causes hepatomegaly, steatohepatitis, and hypertriglyceridemia.
Joshi M, et al.
European Journal of Human Genetics, 22(10), 1229-1229 (2014)
Structure of glycerol-3-phosphate dehydrogenase (GPD1) from Saccharomyces cerevisiae at 2.45 ? resolution.
Alarcon D A, et al.
Acta Crystallographica Section F, Structural Biology and Crystallization Communications, 68(11), 1279-1283 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service